Skip to Content
Merck

Skip To

5135

SpongeCol®

Columnar Architecture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 ea
Check Cart for Availability
€ 604,00
5 ea
Check Cart for Availability
€ 685,00

About This Item

NACRES:
NA.75
UNSPSC Code:
12352207
Pore size:
200 μm average pore size
Diam.:
21 mm , 5135-5EA, 4 mm , 5135-25EA
Sterility:
irradiated, sterile

€ 604,00


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine

Quality Segment

sterility

irradiated, sterile

diam.

21 mm , 5135-5EA, 4 mm , 5135-25EA

thickness

1.5 mm , 5135-5EA

pore size

200 μm average pore size

application(s)

3D cell culture

General description

SpongeCol® is a collagen sponge with an interpenetrating, columnar porous architecture structure. SpongeCol® contains a unique columnar porous network which permits cells and nutrients to flow completely through the interpenetrating pores and provides an increased surface area for cell attachment, growth and migration. It is composed of highly purified Type I collagen which best supports the attachment, proliferation, and function of cells. The collagen is lightly cross-linked for increased mechanical strength and durability for short and long term tissue culture yet is still biodegradable over the longer-term. The diameter of the pores ranges from 100 to 400 micron with the average diameter being approximately 200 microns. This product is terminally sterilized and ready-to-use.

Other Notes

5135-25EA is 4 mm in diameter and 1.5 mm thick and will fit into a 96-well culture plate well. Each package contains 25 collagen sponge discs.

5135-5EA is 21 mm in diameter and 1.5 mm thick and fit into a 12 well culture plate or sanitary luer connectors for flow perfusion. Each package contains 5 collagen sponge discs.

Directions for Use

Preparation and Cell Seeding

Note: Cell attachment to the sponge is generally the most critical step in tissue culture. Temperature, pH, gas exchange and cell concentration can affect the rate and efficiency of attachment. Optimum seeding rate depends on the type of cell being cultured.
1. Aseptically remove the sponge discs from the packaging in a laminar flow work station.
2. Carefully place the sponges into the wells of a 12-well tissue culture plate using a sterile instrument. Be careful not to damage the sponge as it is being transferred. It is recommended to use non-treated tissue culture plasticware.
Note: Tissue-coated plasticware may need to be coated with agarose to prevent cell attachment to the plastic instead of the sponge.
3. Suspend cells at desired concentration (1x104 – 1x105 cells/mL) and dispense sufficient volume of cell solution on top of the sponge placed in the well.
Note: An alternative method is to suspend cells in a neutralized collagen solution (such as PureCol™ type I collagen Catalog #5006 or PureCol™ EZ Gel Catalog #5074). Dispense collagen/cell solution on top of the sponge placed in the well.
4. Transfer to a 37°C incubator for about 1-2 hours to allow for initial cell attachment.
Note: If using the collagen suspension method, the collagen will polymerize at 37°C and encapsulate your cells within the collagen matrix and the sponge.
5. After 1-2 hour, remove the plate from the incubator and check for cell attachment. Additional testing may be required to optimize the time it takes for the cells to attach to the sponge. Check the morphology of the cells. Cell adherence and spreading will dictate the time for attachment.
6. Once the cells have adequately attached to the sponge, increase the final volume in each well to fully cover and provide adequate medium for the culture system.

Changing the Media
1. Change the media 12-24 hours after the initial seeding. The frequency of changes will be determined by cell type, cell attachment efficiency, pH (maintain at pH 7.0 to 7.4) utilization of medium nutrients available to cultures. More frequent medium changes may be required compared to 2D culture systems.

Harvesting of Cells
Note: Protease digestion is the standard method of releasing cells from the sponges. The strength of the attachment of the cells to the collagen sponges will vary from cell line to cell line. The enzyme concentration and digestion time will vary depending upon the activity of the enzyme and the confluence of the cells. Collagenase and/or trypsin may be the preferred method.
1. Washing the sponge with EDTA-PBS may assist the protease digestion. Add sufficient volume to cover the sponge.
2. Aspirate the EDTA-PBS solution from the well.
3. Add sufficient dissociation solution to the well to fully over the sponge.
4. Transfer to a 37°C incubator. Check for cell detachment periodically for cell detachment.
5. Once the cells have fully detached, remove the cells and dispense in a centrifuge tube.
6. Centrifuge the cells as require.

Legal Information

PureCol is a trademark of Advanced BioMatrix, Inc.
SpongeCol is a registered trademark of Advanced BioMatrix, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Z741056Z741050Z741048
description

Columnar Architecture

description

6 mm, pack of 96 ea (supplied in vial)

description

9 mm, pack of 8 ea (supplied in plate)

description

6 mm, pack of 48 ea (supplied in plate)

pore size

200 μm average pore size

pore size

-

pore size

-

pore size

-

diam.

4 mm , 5135-25EA, 21 mm , 5135-5EA

diam.

-

diam.

-

diam.

-

sterility

irradiated, sterile

sterility

-

sterility

-

sterility

-

biological source

bovine

biological source

-

biological source

-

biological source

-

Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

-

thickness

1.5 mm , 5135-5EA

thickness

1 mm

thickness

1 mm

thickness

1 mm


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. What type of collagen is specifically present on this sponge?

    1 answer
    1. This is a product of Advanced BioMatrix, Inc. This product is composed of highly purified Type I collagen.

      Helpful?

Reviews

No rating value

Active Filters