Skip to Content
Merck
All Photos(1)

Documents

51035

Sigma-Aldrich

Fluorescent orange 547

suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H35N2NaO8S2
Molecular Weight:
638.73
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥90% (HPCE)

form

solid

fluorescence

λex 548 nm; λem 567 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

[Na+].CCN1c2ccc(cc2C(C)(CCCC(O)=O)/C1=C\C=C\C3=[N+](CC)c4ccc(cc4C3(C)C)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C30H36N2O8S2.Na/c1-6-31-24-15-13-20(41(35,36)37)18-22(24)29(3,4)26(31)10-8-11-27-30(5,17-9-12-28(33)34)23-19-21(42(38,39)40)14-16-25(23)32(27)7-2;/h8,10-11,13-16,18-19H,6-7,9,12,17H2,1-5H3,(H2-,33,34,35,36,37,38,39,40);/q;+1/p-1

InChI key

CKNWOULQMNPCPW-UHFFFAOYSA-M

Application

Fluorescent orange 547 is a fluorescent biolabel designed for excitation by He/Ne lasers (at 544 nm) or mercury arc lamps. Fluorescent orange 547 reactive exhibits a strong solid state emission. With a negative over all charge Fluorescent orange 547 offers improved water solubility.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service