Skip to Content
Merck
All Photos(1)

Documents

50359

Sigma-Aldrich

Voglibose

≥97.0% (TLC)

Synonym(s):

3,4-Dideoxy-4-{[2-hydroxy-1-(hydroxymethyl)ethyl]amino]-2-D-epi-inositol, N-(1,3-Dihydroxyprop-2-yl)valiolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H21NO7
CAS Number:
Molecular Weight:
267.28
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Assay

≥97.0% (TLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

OCC(CO)N[C@H]1C[C@](O)(CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C10H21NO7/c12-2-5(3-13)11-6-1-10(18,4-14)9(17)8(16)7(6)15/h5-9,11-18H,1-4H2/t6-,7-,8+,9-,10-/m0/s1

InChI key

FZNCGRZWXLXZSZ-CIQUZCHMSA-N

Gene Information

Application

Voglibose is an α-glucosidase inhibitor, similar to acarbose and miglitol, used for lowering post-prandial hyperglycemia (PPHG) in people with diabetes mellitus. Voglibose is used to study it benefits as a protectant against ischemia-reperfusion injury through glucagon-like peptide 1 receptors and the phosphoinositide 3-kinase-Akt-endothelial nitric oxide synthase pathway.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Kawamori et al.
Diabetes, obesity & metabolism, 14(4), 348-357 (2011-12-08)
To evaluate the efficacy and safety of linagliptin 5 and 10 mg vs. placebo and voglibose in Japanese patients with type 2 diabetes mellitus (T2DM). This study enrolled patients with inadequately controlled T2DM who were previously treated with one or
Y Iwamoto et al.
Diabetes, obesity & metabolism, 12(8), 700-708 (2010-07-02)
To confirm the efficacy of vildagliptin in patients with type 2 diabetes (T2D) by testing the hypothesis that glycosylated haemoglobin (HbA1c) reduction with vildagliptin is superior to that with voglibose after 12 weeks of treatment. In this 12-week, randomized, double-blind
Xiaolong Chen et al.
Current medicinal chemistry, 13(1), 109-116 (2006-02-07)
The number of people with diabetes is expected to rise from the current estimated 150 million to 220 million in 2010 and 300 million in 2025, and 90% is Type 2 diabetes or non-insulin dependent diabetes mellitus (NIDDM). Voglibose, one
Y Iwamoto et al.
Diabetes, obesity & metabolism, 12(7), 613-622 (2010-07-02)
To compare the efficacy and safety of sitagliptin (a dipeptidyl peptidase-4 inhibitor) and voglibose (an alpha-glucosidase inhibitor) monotherapy in Japanese patients with type 2 diabetes who have inadequate glycaemic control (HbA1c > or =6.5% and <10.0%) on diet and exercise.
Yutaka Seino et al.
Current medical research and opinion, 27 Suppl 3, 21-29 (2011-12-14)
To compare the efficacy and safety of alogliptin and placebo as add-on therapy in Japanese patients with type 2 diabetes who experienced inadequate glycemic control on voglibose plus diet/exercise therapy. During an 8 week screening phase, patients aged ≥ 20

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service