Skip to Content
Merck

Skip To

50187

Ammonium 2-(methylthio)ethanesulfonate

≥97.0% (TLC)

Synonym(s):

Methyl coenzyme M ammonium salt

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C3H11NO3S2
CAS Number:
Molecular Weight:
173.25
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352106
MDL number:


Quality Level

assay

≥97.0% (TLC)

SMILES string

N.CSCCS(O)(=O)=O

InChI

1S/C3H8O3S2.H3N/c1-7-2-3-8(4,5)6;/h2-3H2,1H3,(H,4,5,6);1H3

InChI key

FSGSBLDVKBYXIK-UHFFFAOYSA-N

Application

Ammonium 2-(methylthio)ethanesulfonate (Methyl coenzyme M) is converted into methane by the enzyme Methyl-coenzyme M reductase (MCR) derived from methanogenic archaea. Methy-coenzyme M is used in studies on methanogenic (methane-producing) enzymatic processes.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



J T Keltjens et al.
European journal of biochemistry, 172(2), 471-476 (1988-03-01)
Cell-free extracts of Methanobacterium thermoautotrophicum (strain delta H) were found to contain high concentrations of inorganic pyrophosphate (up to 40 mM). The compound was accumulated by the organism despite high activity of inorganic pyrophosphatase which was found to be present
K D Olson et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(10), 4099-4103 (1991-05-15)
Inactive 2-(methylthio)ethanesulfonic acid (CH3-S-CoM) reductase was partially activated by exposure to light. This simplified system replaces the complex enzymatic system of protein components A2, A3a, A3b, and ATP, which previously represented the only available means of reactivating the enzyme. Components
Silvan Scheller et al.
Nature, 465(7298), 606-608 (2010-06-04)
Large amounts (estimates range from 70 Tg per year to 300 Tg per year) of the potent greenhouse gas methane are oxidized to carbon dioxide in marine sediments by communities of methanotrophic archaea and sulphate-reducing bacteria, and thus are prevented



Global Trade Item Number

SKUGTIN
50187-10MG04061832860282
50187-50MG04061832860299

Questions

Reviews

No rating value

Active Filters