Skip to Content
Merck

Skip To

49525

L-Glutamic acid γ-(3-carboxy-4-nitroanilide) ammonium salt

≥99.0% (TLC), gamma-glutamyltransferase substrate, powder

Synonym(s):

L-Glutamic acid 5-(3-carboxy-4-nitroanilide) ammonium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Check Cart for Availability
€ 465,00

About This Item

Linear Formula:
C12H12N3O7NH4
CAS Number:
Molecular Weight:
328.28
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352204
EC Number:
264-418-7
MDL number:

€ 465,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

L-Glutamic acid γ-(3-carboxy-4-nitroanilide) ammonium salt, ≥99.0% (TLC)

Quality Segment

assay

≥99.0% (TLC)

form

powder

impurities

≤6% water

solubility

H2O: 100 mg/mL, clear, yellow-green

suitability

suitable as substrate for γ-glutamyltranspeptidase test

storage temp.

2-8°C

SMILES string

N.N[C@@H](CCC(=O)Nc1ccc(c(c1)C(O)=O)[N+]([O-])=O)C(O)=O

InChI

1S/C12H13N3O7.H3N/c13-8(12(19)20)2-4-10(16)14-6-1-3-9(15(21)22)7(5-6)11(17)18;/h1,3,5,8H,2,4,13H2,(H,14,16)(H,17,18)(H,19,20);1H3/t8-;/m0./s1

InChI key

GFABNNMTRVBLPZ-QRPNPIFTSA-N

General description

Substrate for γ-glutamyl transferase.

Application

L-Glutamic acid γ-(3-carboxy-4-nitroanilide) ammonium salt has been used as a synthetic substrate for gamma-glutamyltransferase (GGT) to determine GGT activity.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G613373677G1135
assay

≥99.0% (TLC)

assay

≥98% (HPLC)

assay

≥99.0% (TLC)

assay

≥98% (HPLC)

solubility

H2O: 100 mg/mL, clear, yellow-green

solubility

water: 5 mg/mL, clear, yellow

solubility

H2O: 0.1 g/mL, clear, faintly yellow

solubility

formic acid: 50 mg/mL, clear to slightly hazy

suitability

suitable as substrate for γ-glutamyltranspeptidase test

suitability

-

suitability

suitable as substrate for β-glucuronidase test

suitability

-

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

200

form

powder

form

powder

form

powder or crystals

form

powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters