Skip to Content
Merck
All Photos(1)

Key Documents

48756

Sigma-Aldrich

Genistin

≥97.5% (TLC)

Synonym(s):

4′,5,7-Trihydroxyisoflavone 7-glucoside, Genistein 7-glucoside, Genistein-7-O-β-D-glucopyranoside, Genistoside, NSC 5112

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O10
CAS Number:
Molecular Weight:
432.38
Beilstein:
64479
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97.5% (TLC)

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2cc(O)c3C(=O)C(=COc3c2)c4ccc(O)cc4)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C21H20O10/c22-7-15-18(26)19(27)20(28)21(31-15)30-11-5-13(24)16-14(6-11)29-8-12(17(16)25)9-1-3-10(23)4-2-9/h1-6,8,15,18-24,26-28H,7H2/t15-,18-,19+,20-,21-/m1/s1

InChI key

ZCOLJUOHXJRHDI-CMWLGVBASA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Selective inhibitor of mammalian terminal deoxynucleotidyl transferase (TdT), with no measurable effect on mammalian or microbial DNA polymerases.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bo Yuan et al.
Journal of agricultural and food chemistry, 60(6), 1428-1436 (2012-01-20)
The chemical forms in which isoflavones appear in food or supplements seem to play an important role in their absorption efficiency. However, the influence of the chemical form of isoflavones on their plasma disposition has never been reported, although the
Amélia P Rauter et al.
Phytotherapy research : PTR, 24 Suppl 2, S133-S138 (2010-03-24)
The antihyperglycaemic effect of eight standard flavonoids, previously identified in the ethanol extract of the claimed antidiabetic plant Genista tenera, was evaluated on streptozotocin (STZ)-induced diabetic Wistar rats. The aglycones apigenin, chrysoeriol and genistein, the monoglucosides apigenin 7-O-glucoside, luteolin 7-O-glucoside
Vladimir Ajdžanović et al.
The Journal of membrane biology, 239(3), 131-135 (2010-12-07)
A decrease of erythrocyte membrane fluidity can contribute to the pathophysiology of hypertension. Soy products, which are used as alternative therapeutics in some cardiovascular conditions, contain various isoflavones (genistein, daidzein, and their glucosides, genistin and daidzin), which can incorporate cellular
Anna Hsu et al.
Experimental biology and medicine (Maywood, N.J.), 235(1), 90-97 (2010-04-21)
Previous studies have suggested that soy isoflavones exert anticarcinogenic effects against prostate cancer. We propose that soy extracts, containing a mixture of soy isoflavones and other bioactive components, would be a more potent chemo-preventive agent than individual soy isoflavones. We
Soha M Hamdy et al.
Toxicology and industrial health, 28(8), 746-757 (2011-11-18)
Breast cancer is the second leading cause of cancer death among women and the third most common cancer. In this study, we investigated the chemoprevention efficacy of each of soy genistin, selenium or a combination of them against breast cancer.

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. What is the lambda max for genistin and what is the molar extinction coefficient at that wavelength?

    1 answer
    1. We have data from our QC labs here at Sigma on one of the Genistin products we offer, G0897. The solvent is 85% ethanol/15% water, and the lambda max is typically 262.0 or 262.5 nm. We measured the molar extinction coefficients at the lambda max for four different lots of G0897 back in the 1990s, and the average value was 39,800 M-1.Genistin is a derivative of Genistein. There is an entry for Genistein in the chemicals encyclopedia published by the Royal Society of Chemistry, with information for Genistin under that heading. According to the chemicals encyclopedia published by the Royal Society of Chemistry, 14th edition, entry #4391, uv max (85% ethanol) 262.5 nm (a 90.5)".The value of ""a 90.5"" is the same as an E0.1%, or the extinction coefficient for a 1 mg/mL (or 1 gram per Liter) solution.If one multiplies 90.5 (L/g) by the molecular weight of genistin (432.38 g/mole), you get 39,130 M-1. This is pretty close to the value of 39,800 from our data above."

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service