Skip to Content
Merck

Skip To

44023

2-Acetamido-2-deoxy-D-glucono-1,5-lactone

Synonym(s):

2-(Acetylamino)-2-deoxy-D-gluconic acid δ-lactone, 2-Acetamido-2-deoxy-D-glucono-δ-lactone, N-Acetylglucosamino-1,5-lactone

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C8H13NO6
CAS Number:
Molecular Weight:
219.19
Beilstein/REAXYS Number:
1685090
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
Assay:
≥90.0% (HPLC)


assay

≥90.0% (HPLC)

storage temp.

−20°C

SMILES string

O=C1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1NC(C)=O

InChI

1S/C8H13NO6/c1-3(11)9-5-7(13)6(12)4(2-10)15-8(5)14/h4-7,10,12-13H,2H2,1H3,(H,9,11)/t4?,5-,6+,7?/m0/s1

InChI key

NELQYZRSPDCGRQ-BJBIKLFWSA-N

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters