Skip to Content
Merck
All Photos(1)

Documents

439R-1

Sigma-Aldrich

Mesothelin (EP140) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP140, monoclonal

description

(For In Vitro Diagnostic Use in Select Regions (See Chart))

form

buffered aqueous solution

species reactivity

human

packaging

bottle of 1.0 mL predilute (439R-17)
bottle of 7.0 mL predilute (439R-18)
vial of 0.1 mL concentrate (439R-14)
vial of 0.5 mL concentrate (439R-15)
vial of 1.0 mL concentrate (439R-16)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... MSLN(10232)

General description

Mesothelin is a 40 kDa glycosyl-phosphatudylinositol-anchored glycoprotein cleaved from a 71kDa precursor protein encoded by the human mesothelin gene. Mesothelin is present on the surface of normal mesothelial cells.1 Although the function of mesothelin is unknown, it is overexpressed in a wide variety of cancers including mesothelioma, pancreatic ductal adenocarcinoma, and ovarian carcinoma.1-6 Mesothelin has proven to be a valuable marker for pancreatic ductal adenocarcinoma due to its strong reactivity in tumor tissue and absence in normal pancreas.1, 3-4 Metastatic renal cell carcinoma can present clinical patterns that mimic primary mesothelioma, pancreatic ductal adenocarcinoma, and ovarian carcinoma.2, 4-6 Mesothelin has demonstrated utility in the differential diagnosis between these primary tumors and corresponding metastatic renal cell carcinoma within the context of an antibody panel.4-6 

Quality


IVD

IVD

IVD

RUO

Linkage

Mesothelin Positive Control Slides, Product No. 439S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nelson G Ordóñez
Human pathology, 35(6), 697-710 (2004-06-10)
Both mesotheliomas and renal cell carcinomas can present a wide variety of morphological patterns. Because of this, renal cell carcinomas that metastasize to the pleura and lung may be confused with mesotheliomas. The aim of the present study was to
Nelson G Ordóñez
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 16(3), 192-197 (2003-03-18)
Mesothelin is a cell surface antigen of unknown function that is strongly expressed in mesothelial cells. Although it was reported in 1992 that immunostaining with the K1 anti-mesothelin antibody could be very useful in distinguishing between epithelioid mesotheliomas and pulmonary
Viviane Gnemmi et al.
Analytical and quantitative cytopathology and histopathology, 35(3), 157-162 (2013-12-19)
To determine whether PAX2 and mesothelin immunohistochemistry add additional diagnostic value in discriminating between pancreatic metastasis of renal clear cell carcinoma (PMRCC) and primary ductal adenocarcinoma of the pancreas (PDAC). We retrospectively collected tissue from PMRCC and PDAC. Eleven cases
Erolcan Sayar et al.
JCI insight, 8(7) (2023-02-24)
Prostate-specific membrane antigen (PSMA) is an important cell surface target in prostate cancer. There are limited data on the heterogeneity of PSMA tissue expression in metastatic castration-resistant prostate cancer (mCRPC). Furthermore, the mechanisms regulating PSMA expression (encoded by the FOLH1
Diagnostic value of cytokeratin 7, CD10 and mesothelin in distinguishing ovarian clear cell carcinoma from metastasis of renal clear cell carcinoma.
X Leroy et al.
Histopathology, 51(6), 874-876 (2007-11-29)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service