Skip to Content
Merck

40105

N,N-Dimethyldecylamine N-oxide solution

BioUltra, 0.1 M in H2O

Synonym(s):

DDAO

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)9N(O)(CH3)2
CAS Number:
Molecular Weight:
201.35
PubChem Substance ID:
UNSPSC Code:
12352200
Beilstein/REAXYS Number:
2352550
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

non-ionic

product line

BioUltra

form

liquid

mol wt

201.35 g/mol

concentration

0.1 M in H2O

technique(s)

protein quantification: suitable

impurities

insoluble matter, passes filter test, ≤0.005% peroxides (as H2O2)

refractive index

n20/D 1.337

pH

7.0-8.5 (25 °C, 0.1 M in H2O)

density

0.996 g/mL at 20 °C

anion traces

chloride (Cl-): ≤50 mg/kg, sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤1 mg/kg, As: ≤0.1 mg/kg, Ba: ≤1 mg/kg, Bi: ≤1 mg/kg, Ca: ≤5 mg/kg, Cd: ≤1 mg/kg, Co: ≤1 mg/kg, Cr: ≤1 mg/kg, Cu: ≤1 mg/kg, Fe: ≤1 mg/kg, K: ≤20 mg/kg, Li: ≤1 mg/kg, Mg: ≤1 mg/kg, Mn: ≤1 mg/kg, Mo: ≤1 mg/kg, Na: ≤20 mg/kg, Ni: ≤1 mg/kg, Pb: ≤1 mg/kg, Sr: ≤1 mg/kg, Zn: ≤1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.05, λ: 280 nm Amax: 0.04

SMILES string

CCCCCCCCCC[N+](C)(C)[O-]

InChI

1S/C12H27NO/c1-4-5-6-7-8-9-10-11-12-13(2,3)14/h4-12H2,1-3H3

InChI key

ZRKZFNZPJKEWPC-UHFFFAOYSA-N



Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library