Skip to Content
Merck

3864

Naphthol AS-BI Phosphoric Acid Solution

Synonym(s):

Naphthol AS-BI Phosphoric Acid Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.47
PubChem Substance ID:
UNSPSC Code:
12352106
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

12.5 mg/mL

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

SMILES string

COc1ccccc1NC(=O)c2cc3cc(Br)ccc3cc2OP(O)(O)=O

InChI

1S/C18H15BrNO6P/c1-25-16-5-3-2-4-15(16)20-18(21)14-9-12-8-13(19)7-6-11(12)10-17(14)26-27(22,23)24/h2-10H,1H3,(H,20,21)(H2,22,23,24)

InChI key

HUXIAXQSTATULQ-UHFFFAOYSA-N

Application

For use in Sigma Acid Phosphatase Procedure No. 386.

Other Notes

Napthol AS-BI phosphoric acid, 12.5 mg/mL in N,N-dimethyl formamide.


Still not finding the right product?


signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

136.4 °F - closed cup

flash_point_c

58 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

68-12-2

CAS No.

68-12-2

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library