Skip to Content
Merck
All Photos(1)

Documents

38371

Sigma-Aldrich

Atto 488 Protein Labeling Kit

BioReagent, suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

product line

BioReagent

Quality Level

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 488 nm; λem 520 nm in 0.1 M phosphate buffer, pH 7.0 (recommended)

suitability

suitable for fluorescence

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This kit contains sufficient amounts of reactive dye, buffers and protein purification sets for performing five labeling reactions (1 mg protein each) and for the subsequent purification of the labeled protein.

Application

Atto fluorescent labels are designed for high sensitivity applications, including single molecule detection. Atto labels have rigid structures that do not show any cis-trans-isomerization. Thus these labels display exceptional intensity with minimal spectral shift on conjugation. Atto 488 Protein Labeling Kit is used to produce Atto 488 labeled proteins via conjugation to primary amine groups.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Pallares Pallares et al.
Food & function, 9(12), 6544-6554 (2018-11-28)
The presence of cell walls entrapping starch granules in common bean cotyledons, prevailing after thermal processing and mechanical disintegration, has been identified as the main reason for their (s)low in vitro starch digestibility. Nevertheless, it is unknown if the role
Sheng Shu et al.
Nature communications, 13(1), 4576-4576 (2022-08-06)
Lipopolysaccharide (LPS) is an essential glycolipid and forms a protective permeability barrier for most Gram-negative bacteria. In E. coli, LPS levels are under feedback control, achieved by FtsH-mediated degradation of LpxC, which catalyzes the first committed step in LPS synthesis.
Daniela C Dieterich
Current opinion in neurobiology, 20(5), 623-630 (2010-07-24)
Fluorescent proteins have revolutionized cell biology and, therefore, our understanding of the complex molecular and cellular mechanisms that wire the brain together and enable its plasticity throughout life. The ability to visualize cell biological processes has inspired the development of
Switching modulation for protein labeling with activatable fluorescent probes.
Kalyan K Sadhu et al.
Chembiochem : a European journal of chemical biology, 12(9), 1299-1308 (2011-06-02)
Lan-Tao Gou et al.
Cell, 180(6), 1212-1227 (2020-03-15)
The paternal genome undergoes a massive exchange of histone with protamine for compaction into sperm during spermiogenesis. Upon fertilization, this process is potently reversed, which is essential for parental genome reprogramming and subsequent activation; however, it remains poorly understood how this

Articles

Protein labeling kits with Atto and Tracy dyes provide easy fluorescent labeling of purified proteins, enzymes, and antibodies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service