Skip to Content
Merck
All Photos(1)

Key Documents

358835

Sigma-Aldrich

Diphenylborinic anhydride

95%

Synonym(s):

Oxybis(diphenylborane), Tetraphenyldiboroxane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2BOB(C6H5)2
CAS Number:
Molecular Weight:
346.04
Beilstein:
2950331
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

Assay

95%

form

solid

mp

135-140 °C (lit.)

fluorescence

λex 366 nm; λem 475 nm (after derivatization)

storage temp.

2-8°C

SMILES string

O(B(c1ccccc1)c2ccccc2)B(c3ccccc3)c4ccccc4

InChI

1S/C24H20B2O/c1-5-13-21(14-6-1)25(22-15-7-2-8-16-22)27-26(23-17-9-3-10-18-23)24-19-11-4-12-20-24/h1-20H

InChI key

SNQFEECGHGUHBK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diphenylborinic anhydride (DPBA) may be used for the separation and determination of α-amino acids by boroxazolidone formation. DPBA, an analogue of the vascular gap junction channel blocker, 2-aminoethoxydiphenyl borate (2-APB), may be use in gap junctions channel studies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yan Yang et al.
Biological & pharmaceutical bulletin, 34(9), 1390-1397 (2011-09-02)
2-Aminoethoxydiphenyl borate (2-APB) has recently been demonstrated to inhibit gap junction (GJ) channels, whereas the underlying mechanisms are still unknown. Using mouse TM₄ Sertoli cell which expresses connexin43 (Cx43), we explored the effects of 2-APB and its analogues on dye-coupling
Ke-Tao Ma et al.
American journal of physiology. Heart and circulatory physiology, 300(1), H335-H346 (2010-11-03)
2-Aminoethoxydiphenyl borate (2-APB) analogs are potentially better vascular gap junction blockers than others widely used, but they remain to be characterized. Using whole cell and intracellular recording techniques, we studied the actions of 2-APB and its potent analog diphenylborinic anhydride
C J Strang et al.
Analytical biochemistry, 178(2), 276-286 (1989-05-01)
Reaction of an alpha-amino acid (alpha-AA) with 1,1-diphenylborinic acid (DPBA) leads to the formation of a kinetically stable adduct at pH 2-5 in which both the alpha-amino and the alpha-carboxyl groups are bound to boron forming a cyclic mixed anhydride

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service