Skip to Content
Merck
All Photos(1)

Key Documents

34885

Sigma-Aldrich

3,3′-Dibutylthiacarbocyanine iodide

for fluorescence, ≥98.0%

Synonym(s):

DiSC4(3)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H29IN2S2
CAS Number:
Molecular Weight:
548.55
Beilstein:
3852791
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

for fluorescence

Assay

≥98.0% (TLC)
≥98.0%

form

liquid

solubility

DMSO: soluble
methanol: soluble

fluorescence

λex 559 nm; λem 604 nm in phosphate buffer/SDS pH 7.0

suitability

suitable for fluorescence

SMILES string

[I-].CCCCN1/C(Sc2ccccc12)=C\C=C\c3sc4ccccc4[n+]3CCCC

InChI

1S/C25H29N2S2.HI/c1-3-5-18-26-20-12-7-9-14-22(20)28-24(26)16-11-17-25-27(19-6-4-2)21-13-8-10-15-23(21)29-25;/h7-17H,3-6,18-19H2,1-2H3;1H/q+1;/p-1

InChI key

IBLJGRBTLKWIGM-UHFFFAOYSA-M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iwona T Myszor et al.
Scientific reports, 9(1), 7114-7114 (2019-05-10)
Aroylated phenylenediamines (APDs) are novel inducers of innate immunity enhancing cathelicidin gene expression in human bronchial epithelial cell lines. Here we present two newly developed APDs and aimed at defining the response and signaling pathways for these compounds with reference

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service