Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
25 g | Check Cart for Availability | € 855,00 |
About This Item
description
anionic
Quality Segment
product line
BioUltra
assay
≥99.0% (NT)
form
powder
optical activity
[α]20/D +44±2°, c = 2% in H2O
mol wt
average mol wt 1200-5000
aggregation number
3-12
impurities
insoluble matter, passes filter test, ≤0.5% cholic acid (TLC), ≤6.1% water
pH
7.5-9.5 (25 °C, 0.1 M in H2O)
CMC
2-6
solubility
H2O: 0.1 M at 20 °C, clear, colorless
anion traces
chloride (Cl-): ≤50 mg/kg, sulfate (SO42-): ≤200 mg/kg
cation traces
Al: ≤20 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, Li: ≤5 mg/kg, Mg: ≤20 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤20 mg/kg, Zn: ≤5 mg/kg
λ
0.1 M in H2O
UV absorption
λ: 260 nm Amax: 0.10, λ: 280 nm Amax: 0.08
HLB
16
SMILES string
O.[Na+].C[C@H](CCC([O-])=O)C1CCC2C3CCC4C[C@H](O)CC[C@]4(C)C3C[C@H](O)[C@]12C
InChI
1S/C24H40O4.Na.H2O/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3;;/h14-21,25-26H,4-13H2,1-3H3,(H,27,28);;1H2/q;+1;/p-1/t14-,15-,16-,17+,18-,19+,20+,21+,23+,24-;;/m1../s1
InChI key
NDVHNZISGVSFMP-WTCAICSISA-M
General description
Application
Biochem/physiol Actions
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥99.0% (NT) | assay ≥99.0% (titration) | assay ≥97% (titration) | assay ≥99% |
| form powder | form powder | form powder | form powder |
| aggregation number 3-12 | aggregation number - | aggregation number 3-12 | aggregation number 2-3 |
| Quality Level 300 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| mol wt average mol wt 1200-5000 | mol wt 432.57 g/mol | mol wt micellar avg mol wt 1200-5000 | mol wt micellar avg mol wt 900-1300 |
| solubility H2O: 0.1 M at 20 °C, clear, colorless | solubility H2O: 0.1 M, clear, colorless | solubility water: 50 mg/mL, clear, colorless to light yellow | solubility H2O: 1 M, clear, faintly yellow to yellow |
Still not finding the right product?
Explore all of our products under Sodium deoxycholate monohydrate
signalword
Warning
hcodes
pcodes
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



