Skip to Content
Merck
All Photos(1)

Key Documents

30382

Sigma-Aldrich

Cytochalasin C from Metarrhizium anisopliae

≥97.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H37NO6
CAS Number:
Molecular Weight:
507.62
Beilstein:
1613194
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

Metarrhizium anisopliae

Assay

≥97.0% (TLC)

form

fibers
powder

solubility

methylene chloride: 5 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

C[C@@H]1C\C=C\[C@H]2[C@H](O)C(C)=C(C)C3[C@H](Cc4ccccc4)NC(=O)[C@@]23C(OC(C)=O)\C=C\[C@@](C)(O)C1=O

InChI

1S/C30H37NO6/c1-17-10-9-13-22-26(33)19(3)18(2)25-23(16-21-11-7-6-8-12-21)31-28(35)30(22,25)24(37-20(4)32)14-15-29(5,36)27(17)34/h6-9,11-15,17,22-26,33,36H,10,16H2,1-5H3,(H,31,35)/b13-9+,15-14+/t17-,22+,23+,24-,25?,26-,29-,30-/m1/s1

InChI key

NAIODHJWOHMDJX-QMTXKCDBSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Structure of cytochalasins and cytochalasin B binding sites in human erythrocyte membranes

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Repr. 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong-Xiang Song et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 33(6), 901-903 (2010-11-06)
To study the secondary metabolites of mangrove endophytic fungus BL321. The compounds were isolated by chromatographic technique. The structures were identified by comprehensive physico-chemical properties and spectral methods. Five compounds were isolated and identified as 3,4a-dimethyl-2-oxo-2,4,4a,5,6,7-hexahydronaphtho[2,3-b]furan-5-carboxylic acid(1), cytochalasin C(2), cytochalasin
T Glinsukon et al.
Research communications in chemical pathology and pharmacology, 51(2), 265-268 (1986-02-01)
Cytochalasins A, B, C, D and E at a concentration of 5.0 microgram/ml significantly inhibited the motility of spermatozoa collected from the cauda epididymidis of rats when diluted in Hank's solution containing BSA by 21.6-38.1% within a few min incubation.
H Haimoto et al.
The American journal of pathology, 121(2), 185-191 (1985-11-01)
To elucidate the changes of ultrastructural localization of S-100 protein during lipolysis in adipocytes, an immunoelectron-microscopic study was performed. Epididymal fat pads from Wistar rats were incubated in the buffer with or without 10 microM epinephrine. Before incubation with epinephrine
V G Lukashin et al.
Morfologiia (Saint Petersburg, Russia), 118(4), 41-45 (2003-03-13)
Colchicine and cytochalasine C exert modifying action on tissue receptors of urinary bladder of the frog. Under the effect of cytochalasine the period of vital staining of receptors were changed. Latent period of decoloration grew 37-56% longer while the period
E A Walling et al.
Archives of biochemistry and biophysics, 264(1), 321-332 (1988-07-01)
The effects on actin self-assembly of 9 common cytochalasins and 9 synthetic analogs have been assayed using fluorescence photobleaching recovery (FPR). The specific assembly activities of cytochalasins determined by this assay are (i) reduction of the fraction of actin molecules

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service