Skip to Content
Merck
All Photos(1)

Key Documents

30312

Sigma-Aldrich

Nα-Methyl-L-lysine monohydrochloride

≥98.0% (TLC)

Synonym(s):

(S)-6-Amino-2-(methylamino)hexanoic acid hydrochloride, N-Me-Lys-OH · HCl

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€226.00
500 MG
€891.00

€226.00


Estimated to ship on19 May 2025



Select a Size

Change View
100 MG
€226.00
500 MG
€891.00

About This Item

Empirical Formula (Hill Notation):
C7H16N2O2 · HCl
CAS Number:
Molecular Weight:
196.68
Beilstein:
6118109
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

€226.00


Estimated to ship on19 May 2025


Quality Level

Assay

≥98.0% (TLC)

optical activity

[α]/D 27.5±1.5°, c = 0.1 in 1 M HCl

SMILES string

Cl.CN[C@@H](CCCCN)C(O)=O

InChI

1S/C7H16N2O2.ClH/c1-9-6(7(10)11)4-2-3-5-8;/h6,9H,2-5,8H2,1H3,(H,10,11);1H/t6-;/m0./s1

InChI key

LWUKQYOOMILVOV-RGMNGODLSA-N

Biochem/physiol Actions

Nα-Methyl-L-lysine was shown to inhibit the growth and sporulation of Penicillium chrysogenum. [1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C G Friedrich et al.
Applied and environmental microbiology, 34(6), 706-709 (1977-12-01)
Compounds structurally related to lysine were tested against Penicillium chrysogenum Wis. 54-1255 for inhibition of growth, sporulation, and penicillin formation. This strain is relatively resistant to lysine analogs. The compounds that were the more active inhibitors of growth and whose
D J Gordon et al.
Biochemistry, 40(28), 8237-8245 (2001-07-11)
A potential goal in the prevention or therapy of Alzheimer's disease is to decrease or eliminate neuritic plaques composed of fibrillar beta-amyloid (Abeta). In this paper we describe N-methyl amino acid containing congeners of the hydrophobic "core domain" of Abeta

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service