Skip to Content
Merck
All Photos(1)

Documents

3014

Sigma-Aldrich

Transcreener® GDP FI Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122100
NACRES:
NA.81

shipped in

dry ice

storage temp.

−20°C

General description

The Transcreener® GDP FI Assay is a competitive immunoassay for GDP with a red fluorescent intensity (FI) readout. Designed specifically for HTS and featuring a single addition mix-and-read format and room temperature reagent stability, the Transcreener® GDP FI Assay is compatible with plate readers typically found in research labs as well as those used in HTS core facilities. The 3014-A kit is formatted for 96-well plates while the 3014-1K and 3014- 10K kits are for 384-well and higher densities.

Able to accommodate GTP concentrations ranging from 0.1 to 1,000 μM, the Transcreener® GDP FI Assay provides excellent data quality (Z′ = 0.7) at low substrate conversion (typically 10% or less). Because the proprietary antibody used is highly selective for GDP, the assay can be used with any enzyme that converts GTP to GDP, including small G proteins. (Note that many commercially available GTPase preparations contain GDP as a stabilizer, which must be removed or diluted prior to use with this assay so as not to contribute to background signal.

View full Transcreener® product list

Quantity

3014-1K = 1,000 assay, 384-well
3014-10K = 10,000 assay, 384-well
3014-A = 200 assay, 96-well

Physical form

Kit with buffered aqueous solutions

Legal Information

Transcreener is a registered trademark of BellBrook Labs

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Won Kang et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 7(24), 2002142-2002142 (2020-12-22)
NH3, essential for producing artificial fertilizers and several military and commercial products, is being produced at a large scale to satisfy increasing demands. The inevitable leakage of NH3 during its utilization, even in trace concentrations, poses significant environmental and health
Mengyang Zhang et al.
Oxidative medicine and cellular longevity, 2021, 8877540-8877540 (2021-05-20)
The aim of the present study was to investigate the protective effects and mechanisms of KLK1 on aging-related prostate alterations and search clues about the application of KLK1 to the treatment of human BPH. Thirty-six rats including 26 male wild-type
Gaurav Kumar et al.
Bio-protocol, 8(24) (2019-01-11)
Macrophages are highly phagocytic cells that utilize various pathogen recognition receptors (PRRs) to recognize pathogen-associated molecular patterns (PAMPs). These PAMPs can be present within the microbe, such as bacterial CpG DNA, and are recognized by Toll-like receptor 9 (TLR9), a

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service