Skip to Content
Merck
All Photos(1)

Documents

2671R

Millipore

BIOEAZE Rigging Sets

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103801

description

Designed for use with the SAFC Biosciences' QUICK SCALE™ Program to Allow for Multiple Applications

sterility

sterile; γ-irradiated

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

3/16" x 3/8" Silicone Tubing with Female Quick Connect on One End and Filling Bell with Cap on One End

Application

Transfer set for dispensing with a filling bell into bottles

Packaging

Double Polybagged

Legal Information

BIOEAZE is a trademark of Sigma-Aldrich Co. LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service