Skip to Content
Merck
All Photos(1)

Key Documents

21965

Sigma-Aldrich

5(6)-Carboxy-X-rhodamine

BioReagent, suitable for fluorescence

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€191.00
100 MG
€606.00

€191.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€191.00
100 MG
€606.00

About This Item

Empirical Formula (Hill Notation):
C33H30N2O5
CAS Number:
Molecular Weight:
534.60
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.32

€191.00


Estimated to ship on30 May 2025


Request a Bulk Order

product line

BioReagent

mp

≥250 °C (lit.)

fluorescence

λex 570 nm; λem ~595 nm in 0.1 M Tris pH 8.0

suitability

suitable for fluorescence

SMILES string

OC(=O)c1ccc2c(c1)C(=O)OC23c4cc5CCCN6CCCc(c4Oc7c8CCCN9CCCc(cc37)c89)c56.OC(=O)c%10ccc%11C(=O)OC%12(c%13cc%14CCCN%15CCCc(c%13Oc%16c%17CCCN%18CCCc(cc%12%16)c%17%18)c%14%15)c%11c%10

InChI

1S/2C33H30N2O5/c36-31(37)20-9-10-24-23(15-20)32(38)40-33(24)25-16-18-5-1-11-34-13-3-7-21(27(18)34)29(25)39-30-22-8-4-14-35-12-2-6-19(28(22)35)17-26(30)33;36-31(37)20-9-10-21-24(17-20)33(40-32(21)38)25-15-18-5-1-11-34-13-3-7-22(27(18)34)29(25)39-30-23-8-4-14-35-12-2-6-19(28(23)35)16-26(30)33/h2*9-10,15-17H,1-8,11-14H2,(H,36,37)

InChI key

KLNOCCPJAOCRHF-UHFFFAOYSA-N

Application

5(6)-Carboxy-X-rhodamine may be used as a fluorescent dye in nucleic acid analytical research applications.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

partially soluble in methanol, 0.1 M Tris pH 8.0

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jin-Oh You et al.
International journal of nanomedicine, 1(2), 173-180 (2007-08-29)
Reverse microemulsion was used as a template to fabricate chitosan-alginate core-shell nanoparticles encapsulated with enhanced green fluorescent protein (EGFP)-encoded plasmids. The average size of DNA-entrapped nanoparticles measured by dynamic light scattering was increased proportionally, with the N/P ratios ranging from
Gadi Cohen et al.
Arteriosclerosis, thrombosis, and vascular biology, 39(8), 1602-1613 (2019-06-14)
The early embryo implantation is characterized by enhanced uterine vascular permeability at the site of blastocyst attachment, followed by extracellular-matrix remodeling and angiogenesis. Two TG (transglutaminase) isoenzymes, TG2 (tissue TG) and FXIII (factor XIII), catalyze covalent cross-linking of the extracellular-matrix.
Tae Seok Seo et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(17), 5926-5931 (2005-04-15)
We report four-color DNA sequencing by synthesis (SBS) on a chip, using four photocleavable fluorescent nucleotide analogues (dGTP-PC-Bodipy-FL-510, dUTP-PC-R6G, dATP-PC-ROX, and dCTP-PC-Bodipy-650) (PC, photocleavable; Bodipy, 4,4-difluoro-4-bora-3alpha,4alpha-diaza-s-indacene; ROX, 6-carboxy-X-rhodamine; R6G, 6-carboxyrhodamine-6G). Each nucleotide analogue consists of a different fluorophore attached to
Jun Wang et al.
Clinical chemistry, 56(4), 623-632 (2010-03-09)
The EGFR [epidermal growth factor receptor (erythroblastic leukemia viral (v-erb-b) oncogene homolog, avian)] gene is known to harbor genomic alterations in advanced lung cancer involving gene amplification and kinase mutations that predict the clinical response to EGFR-targeted inhibitors. Methods for
Silvia Mannucci et al.
International journal of nanomedicine, 15, 1745-1758 (2020-03-28)
Solid lipid nanoparticles are largely used in biomedical research and are characterized by high stability and biocompatibility and are also able to improve the stability of various loaded molecules. In vitro studies demonstrated that these nanoparticles are low cytotoxic, while

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service