Skip to Content
Merck
All Photos(1)

Documents

210-500

Sigma-Aldrich

Endothelial Cell Basal Medium (500ml)

Synonym(s):

Primary Cell Culture Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

form

liquid

Quality Level

shelf life

1 yr

manufacturer/tradename

Cell Applications, Inc

shipped in

dry ice

storage temp.

−20°C

General description

Basal Medium without Growth Supplement

Application

Endothelial Cell Basal Medium (500ml) has been used in culturing of:
  • human umbilical vein endothelial cells
  • human coronary artery endothelial cells
  • human pulmonary artery endothelial cells

Quality

Each lot was tested for the ability to support plating, spreading and proliferation

Preparation Note

contained in protocol

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richa Pande et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 140, 111788-111788 (2021-06-02)
Complex regional pain syndrome (CRPS) is a chronic pain condition characterized by inflammation and debilitating pain. CRPS patients with pain refractory to more conventional analgesics can be treated with subanesthetic doses of ketamine. Our previous studies found that poor responders
Lian-Yu Lin et al.
The Science of the total environment, 414, 68-72 (2011-11-23)
To pray in temples is a regular activity in Buddhism and Taoism societies, yet few studies investigated the effects of particles from incense-burning in temples. The objectives of this study are to examine particle size and polycyclic aromatic hydrocarbon (PAH)
Liza Merly et al.
Immunopharmacology and immunotoxicology, 39(2), 55-58 (2017-02-06)
The popular murine macrophage cell line, RAW 264.7, is often used to initially screen natural products for bioactivity and to predict their potential effect in vivo or on primary cells. The cell line response is considered to reflect the potential
Daria Shishkova et al.
International journal of molecular sciences, 21(21) (2020-11-01)
Although saphenous veins (SVs) are commonly used as conduits for coronary artery bypass grafting (CABG), internal thoracic artery (ITA) grafts have significantly higher long-term patency. As SVs and ITA endothelial cells (ECs) have a considerable level of heterogeneity, we suggested
Hong Xue et al.
International journal of chronic obstructive pulmonary disease, 16, 1329-1345 (2021-05-20)
Etanercept (ETN), a tumor necrosis factor-α (TNF-α) inhibitor, has been applied in the treatment of many diseases. However, whether it has effects on chronic obstructive pulmonary disease (COPD) and its interaction with tumor necrosis factor receptor 1 (TNFR1) remained unknown.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service