Skip to Content
Merck
All Photos(1)

Documents

208R-1

Sigma-Aldrich

GH (EP267) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP267, monoclonal

description

(For In Vitro Diagnostic Use in Select Regions (See Chart))

form

buffered aqueous solution

species reactivity

human

packaging

bottle of 1.0 mL predilute (208R-17)
bottle of 7.0 mL predilute (208R-18)
vial of 0.1 mL concentrate (208R-14)
vial of 0.5 mL concentrate (208R-15)
vial of 1.0 mL concentrate (208R-16)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

Gene Information

mouse ... Gh(14599)

General description

Pituitary growth hormone (GH) plays a crucial role in stimulating and controlling the growth, metabolism and differentiation of many mammalian cell types by modulating the synthesis of multiple mRNA species. These effects are mediated by the binding of GH to its membrane-bound receptor, GHR, and involve a phosphorylation cascade that results in the modulation of numerous signaling pathways. GH is synthesized by acidophilic or somatotropic cells of the anterior pituitary gland. Human growth hormone contains 191 amino acid residues with two disulfide bridges. Anti-GH is a useful marker in classification of pituitary tumors and the study of pituitary disease (acromegaly).1-3 It reacts with GH-producing cells. Growth hormone receptors have been found in various non-pituitary cells, including that from hepatocellular carcinoma and various benign and malignant cutaneous lesions.4-6

Quality


IVD

IVD

IVD

RUO

Linkage

GH Positive Control Slides, Product No. 208S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T García-Caballero et al.
Endocrine, 12(3), 265-271 (2000-08-30)
The liver is an essential target tissue for growth hormone (GH) and prolactin (PRL). The aim of this study was to determine the in situ expression of growth hormone receptor (GHR) and prolactin receptor (PRLR) in hepatocellular carcinomas and to
L K Chopin et al.
Growth hormone & IGF research : official journal of the Growth Hormone Research Society and the International IGF Research Society, 12(2), 126-136 (2002-08-15)
Prostate cancer is a significant cause of morbidity and mortality in Western males. While it is known that androgens play a central role in prostate cancer development and progression, other hormones and growth factors are also involved in prostate growth.
A Matsuno et al.
Pathology, research and practice, 197(1), 13-20 (2001-02-24)
To characterize the morphological and functional aspects of silent somatotroph adenomas with paradoxical responses of GH in TRH or GnRH provocation tests, which are considered to be a useful strategy for endocrinological identification of silent somatotroph adenomas, we examined three
K S G Cunha et al.
Journal of clinical pathology, 56(10), 758-763 (2003-09-30)
The hallmark of neurofibromatosis type 1 (NF1) is the development of multiple neurofibromas. Solitary neurofibroma may occur in an individual who does not have NF1, but multiple neurofibromas tend to develop only in those with NF1. It has been suggested

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service