Skip to Content
Merck
All Photos(1)

Key Documents

19056

Sigma-Aldrich

2-Methoxy-2-(2-naphthyl)acetonitrile

Synonym(s):

α-Methoxy-2-naphthaleneacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11NO
CAS Number:
Molecular Weight:
197.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95.0% (HPLC)

Quality Level

fluorescence

λex 227 nm; λem 336 nm in methanol

SMILES string

COC(C#N)c1ccc2ccccc2c1

InChI

1S/C13H11NO/c1-15-13(9-14)12-7-6-10-4-2-3-5-11(10)8-12/h2-8,13H,1H3

InChI key

JDVRUILJVSKUFH-UHFFFAOYSA-N

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teresa F Reyes et al.
Regulatory toxicology and pharmacology : RTP, 119, 104817-104817 (2020-11-11)
The production of soy leghemoglobin C2 (LegH) by Pichia pastoris (syn. K. phaffii) was developed by Impossible Foods to serve as a sustainable source of flavor and aroma in plant-based meats. The potential allergenicity and toxicity of a LegH from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service