Skip to Content
Merck
All Photos(1)

Documents

18360

Sigma-Aldrich

Bromosulfalein

for liver diagnosis, for microscopy (Hist.)

Synonym(s):

Phenoltetrabromophthalein-3′,3″-disulfonic acid disodium salt, Sulfobromophthalein disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H8Br4Na2O10S2
CAS Number:
Molecular Weight:
838.00
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

grade

for liver diagnosis
for microscopy (Hist.)

SMILES string

[Na+].[Na+].OC(=O)c1c(Br)c(Br)c(Br)c(Br)c1\C(c2ccc(O)c(c2)S([O-])(=O)=O)=C3/C=CC(=O)C(=C3)S([O-])(=O)=O

InChI

1S/C20H10Br4O10S2.2Na/c21-16-14(15(20(27)28)17(22)19(24)18(16)23)13(7-1-3-9(25)11(5-7)35(29,30)31)8-2-4-10(26)12(6-8)36(32,33)34;;/h1-6,25H,(H,27,28)(H,29,30,31)(H,32,33,34);;/q;2*+1/p-2/b13-8+;;

InChI key

HXUITPQMHVLBNV-LIYVDSPJSA-L

Application

Reagent for the quantitative determination of proteins.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen Zhang et al.
European journal of drug metabolism and pharmacokinetics, 42(4), 669-676 (2016-11-07)
Ophiopogonin D (OPD) is one of the main active ingredients of SMI (Shenmai injection) which is widely used in clinical practice in China. Our previous study indicated  that OPD might be transported from blood into liver mediated by organic anion
D Yao et al.
Neuroscience, 227, 283-292 (2012-10-13)
Clinical studies and animal models have shown that pharmacoresistant epilepsy is partly due to the overexpression of ATP-binding cassette transporters at the brain. The purposes of the study were to investigate the function and expression of multidrug resistance-associated protein 2
Cecilia L Balaban et al.
Artificial organs, 35(5), 508-515 (2011-05-21)
The insults sustained by transplanted livers (hepatectomy, hypothermic preservation, and normothermic reperfusion) could compromise hepatic function. Hydrogen sulfide (H₂S) is a physiologic gaseous signaling molecule, like nitric oxide (NO) and carbon monoxide (CO). We examined the effect of diallyl disulfide
Jörg König et al.
Drug metabolism and disposition: the biological fate of chemicals, 39(6), 1097-1102 (2011-03-25)
The therapeutic effects and metabolism of mesalazine (5-aminosalicylic acid) in patients with inflammatory bowel disease require intracellular accumulation of the drug in intestinal epithelial cells and hepatocytes. The molecular mechanisms of mesalazine uptake into cells have not been characterized so
Chen Yanxiao et al.
Life sciences, 89(1-2), 1-6 (2011-04-07)
The purpose of the present study was to investigate the roles of transporters in the renal excretion of entecavir. We analyzed the effect of probenecid, cimetidine, sulfobromophthalein sodium (BSP), verapamil, inhibitors of organic anion transporter (OAT), organic cation transporter (OCT)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service