Skip to Content
Merck

Skip To

17480

Bromocresol Green sodium salt

suitable for microscopy, ACS reagent

Synonym(s):

3′,3″,5′,5″-Tetrabromo-m-cresolsulfonphthalein sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 g
Check Cart for Availability
€ 185,00

About This Item

Empirical Formula (Hill Notation):
C21H13Br4NaO5S
CAS Number:
Molecular Weight:
720.00
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
41116124
EC Number:
263-657-4
MDL number:

€ 185,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent

Quality Segment

mp

230 °C (dec.) (lit.)

suitability

suitable for microscopy

SMILES string

[Na+].Cc1c(Br)c(O)c(Br)cc1\C(=C2\C=C(Br)C(=O)C(Br)=C2C)c3ccccc3S([O-])(=O)=O

InChI

1S/C21H14Br4O5S.Na/c1-9-12(7-14(22)19(26)17(9)24)21(13-8-15(23)20(27)18(25)10(13)2)11-5-3-4-6-16(11)31(28,29)30-21;/h3-8,26-27H,1-2H3;/q;+1/p-1

InChI key

HEFSGAHJDGZCHA-UHFFFAOYSA-M

Application

Bromocresol green has been used to study the removeal of non-ionic organic pollutants from water via liquid-liquid extraction. Bromocresol green has also been used in a study to examine growth inhibition pattern and lactic acid production in Streptococcus mutans.


Still not finding the right product?

Explore all of our products under Bromocresol Green sodium salt


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters