Skip to Content
Merck

Skip To

168024K

Vitronectin Mimetic peptides MAPTRIX-V-KKQRFRHRNRKGY

animal-free ECM peptides, solution, 1.0 mg/mL protein, suitable for cell culture

Synonym(s):

MAPTrix Technology

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Usually ships in 3 weeks.
€ 641,00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.75
Form:
aqueous solution
Technique(s):
cell culture | mammalian: suitable
Concentration:
1.0 mg/mL protein
Mol wt:
~24.7 kDa

€ 641,00


Usually ships in 3 weeks.

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

MAPTRIX-V-KKQRFRHRNRKGY, Vitronectin Mimetic, aqueous solution

sterility

Filtered sterilized solution

form

aqueous solution

mol wt

~24.7 kDa

packaging

pkg of 10 mg

concentration

1.0 mg/mL protein

technique(s)

cell culture | mammalian: suitable

binding specificity

Receptor: Heparin
Domain: HBV
Binding Motif: KKQRFRHRNRKGYRSQ

shipped in

ambient

storage temp.

2-8°C

Application

MAPTrix Technology provides a true extracellular microenvironment (ECM) by presenting combinatorial peptide motifs. The ECM, defined by biochemical cues and physical cues, is a deciding factor in a wide range of cellular processes including cell adhesion, proliferation, differentiation, and expression of phenotype-specific functions.Currently existing technology offers simple and merely adequate environments that facilitate simple cell processes such as cellular adhesion. The simple presentation of cell adhesion motifs is not optimal for controlling more integrated processes. Crosstalk among signaling pathways act synergistically to enhance cellular responses such as cell adhesion or proliferation. A study showed that a combination of extracellular matrix derived peptides presented on a surface may enhance cell adhesion strength and focal adhesion assembly. The combinatorial presentation of ECM peptides on cell growth surfaces may also promote elevated proliferation rates of primary or stem cells.

Features and Benefits

  • Biochemically-defined, animal-free cell culture surfaces designed to enhance cell performance
  • MAPTrix produces a uniform ECM surface that provides a highly controlled 2D extracellular microenvironment for cell cultures and related applications
  • Adhesion properties of the mussel adhesive protein makes the coating reproducible & reliable

Legal Information

MAPTrix is a trademark of Kollodis Biosciences

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
168014K166214K162294K
description

Vitronectin Mimetic, aqueous solution

description

Vitronectin Mimetic, aqueous solution

description

Collagen Mimetic, aqueous solution

description

Laminin Mimetic, aqueous solution

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

sterility

Filtered sterilized solution

sterility

Filtered sterilized solution

sterility

Filtered sterilized solution

sterility

Filtered sterilized solution

concentration

1.0 mg/mL protein

concentration

1.0 mg/mL protein

concentration

1.0 mg/mL protein

concentration

1.0 mg/mL protein

form

aqueous solution

form

aqueous solution

form

aqueous solution

form

aqueous solution

mol wt

~24.7 kDa

mol wt

~23.8 kDa

mol wt

~23.9 kDa

mol wt

~24.2 kDa

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Storage Class

12 - Non Combustible Liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters