Skip to Content
Merck

14951

2-[(1E)-2-(4-Azidophenyl)ethenyl]-6-methoxybenzothiazole

azide clickable fluorogenic marker 453

Synonym(s):

(E)-6-Methoxy-2-(4-azidostyryl)-1,3-benzothiazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H12N4OS
CAS Number:
Molecular Weight:
308.36
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352200
Beilstein/REAXYS Number:
23580408
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95.0% (HPLC)

fluorescence

λex 355 nm; λem 449 nm±10 nm in DMSO

storage temp.

−20°C

SMILES string

COC1=CC=C2C(SC(/C=C/C3=CC=C(N=[N+]=[N-])C=C3)=N2)=C1

InChI

1S/C16H12N4OS/c1-21-13-7-8-14-15(10-13)22-16(18-14)9-4-11-2-5-12(6-3-11)19-20-17/h2-10H,1H3/b9-4+

InChI key

ZOEXSFZBSQSYDV-RUDMXATFSA-N



pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library