Skip to Content
Merck

Skip To

14513

1,2-Bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetic acid tetrasodium salt

≥95% (HPLC)

Synonym(s):

1,2-Bis (2-aminophenoxy) ethane-N,N,N′,N′-tetraacetic acid tetrasodium salt, BAPTA-Na4, Tetrasodium 1,2-bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Check Cart for Availability
€ 257,00

About This Item

Empirical Formula (Hill Notation):
C22H20N2Na4O10
CAS Number:
Molecular Weight:
564.36
UNSPSC Code:
12352106
NACRES:
NA.32
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
8818619
Assay:
≥95% (HPLC)
Form:
powder

€ 257,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

impurities

≤12.5% water

solubility

H2O: 10 mg/mL, clear, faintly yellow

fluorescence

λex 254 nm; λem 363 nm in H2O, λex 274 nm; λem 363 nm (with calcium)

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CC([O-])=O)c1ccccc1OCCOc2ccccc2N(CC([O-])=O)CC([O-])=O

InChI

1S/C22H24N2O10.4Na/c25-19(26)11-23(12-20(27)28)15-5-1-3-7-17(15)33-9-10-34-18-8-4-2-6-16(18)24(13-21(29)30)14-22(31)32;;;;/h1-8H,9-14H2,(H,25,26)(H,27,28)(H,29,30)(H,31,32);;;;/q;4*+1/p-4

InChI key

ZWSMLJACYSGFKD-UHFFFAOYSA-J

Application

BAPTA-Na4 is a fluorescent compound utilized to determine free Ca2+ by performing fluorescence emission scans at an excitation wavelength of 299 nm and measuring the emission wavelength at 360 nm .
suitable as Ca2+-indicator (fluorescence suppressed by Ca2+)

Other Notes

Chelator with a greater selectivity for calcium compared with magnesium (spectrophotometric monitoring):[1]


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters