Skip to Content
Merck
All Photos(1)

Key Documents

13920

Sigma-Aldrich

O-Benzyl-DL-serine

≥99.0% (NT)

Synonym(s):

(±)-2-Amino-3-benzyloxypropionic acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€ 628,30
1 G
€ 1.205,80
2 G
€ 2.188,30
5 G
€ 4.670,80
10 G
€ 8.713,30

Vendor SKU: 5F-010-500MG

€ 628,30


Availability
Ships in 21 days - from Aldrich Partner.

Request a Bulk Order

Select a Size

Change View
500 MG
€ 628,30
1 G
€ 1.205,80
2 G
€ 2.188,30
5 G
€ 4.670,80
10 G
€ 8.713,30

About This Item

Linear Formula:
C6H5CH2OCH2CH(NH2)COOH
CAS Number:
Molecular Weight:
195.22
Beilstein:
3204905
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Vendor SKU: 5F-010-500MG

€ 628,30


Availability
Ships in 21 days - from Aldrich Partner.

Request a Bulk Order

Assay

≥99.0% (NT)

form

solid

mp

~227 °C (dec.)

application(s)

peptide synthesis

SMILES string

NC(COCc1ccccc1)C(O)=O

InChI

1S/C10H13NO3/c11-9(10(12)13)7-14-6-8-4-2-1-3-5-8/h1-5,9H,6-7,11H2,(H,12,13)

InChI key

IDGQXGPQOGUGIX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service