Skip to Content
Merck
All Photos(1)

Key Documents

137276

SAFC

BIS-TRIS

EMPROVE® EVOLVE

Pharma Manufacturing

Synonym(s):

BIS-TRIS, 2,2-Bis(hydroxymethyl)-2,2′,2″-nitrilotriethanol, 2-Bis(2-hydroxyethyl)amino-2-(hydroxymethyl)-1,3-propanediol, Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H19NO5
CAS Number:
Molecular Weight:
209.24
Beilstein:
2205275
MDL number:
UNSPSC Code:
12161705
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

product line

EMPROVE® EVOLVE

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, microbial, and trace metals; tested

useful pH range

5.8-7.2

pKa (25 °C)

6.5

solubility

water: 0.1 M, clear, colorless

suitability

suitable for manufacturing use

storage temp.

2-30°C

SMILES string

OCCN(CCO)C(CO)(CO)CO

InChI

1S/C8H19NO5/c10-3-1-9(2-4-11)8(5-12,6-13)7-14/h10-14H,1-7H2

InChI key

OWMVSZAMULFTJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bis Tris is a commonly used biological buffer for very broad usage in biological systems such as diagnostics, electrophoresis and protein based bioprocessing. The pKa of Bis Tris is 6.36 which makes it a good candidate for biological-based formulations.

Bis Tris is used in a wide variety of biological applications including protein separation buffers, cell culture media, electrophoresis separations and diagnostic reagent formulations.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service