Skip to Content
Merck
All Photos(1)

Key Documents

09416

Sigma-Aldrich

21 L-Amino acids + Glycine

≥99.0% (NT)

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
€580.00

€580.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 SET
€580.00

About This Item

EC Number:
UNSPSC Code:
12352209
eCl@ss:
32160406
NACRES:
NA.26

€580.00


Check Cart for Availability

Request a Bulk Order

Assay

≥99.0% (NT)

impurities

≤0.3% foreign amino acids

Components

1 g of each: L-Alanine, L-Arginine hydrochloride, L-Asparagine, L-Aspartic acid, L-Cysteine, L-Cystine, L-Glutamic acid, L-Glutamine, Glycine, L-Histidine hydrochloride, L-4-Hydroxyproline, L-Isoleucine, L-Leucine, L-Lysine hydrochloride, L-Methionine, L-Phenylalanine, L-Proline, L-Serine, L-Threonine, L-Tryptophan, L-Tyrosine, L-Valine

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What material is the packaging made of? What material is the cap (closures) made of?

    1 answer
    1. All the components of 09416-1EA are assigned the same bottle and cap:
      Narrow neck, round amber glass bottle, 10 ml. (Hydrolytic Class III)
      Red polypropylene screw cap with tamper evident seal and PTFE/EPDM flat cap Liner

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service