Skip to Content
Merck

Skip To

08550

Aminostilbamidine methanesulfonate salt

BioReagent, suitable for fluorescence, ≥97.0% (HPLC)

Synonym(s):

2-Aminostilbene-4,4′-dicarbamidine methanesulfonate salt

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C16H17N5 · CH4O3S
CAS Number:
Molecular Weight:
375.45
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:


product line

BioReagent

assay

≥97.0% (HPLC)

fluorescence

λex 386 nm; λem 540 nm in TE buffer; DNA

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

CS(O)(=O)=O.NC(=N)c1ccc(cc1)\C=C\c2ccc(cc2N)C(N)=N

InChI

1S/C16H17N5.CH4O3S/c17-14-9-13(16(20)21)8-7-11(14)4-1-10-2-5-12(6-3-10)15(18)19;1-5(2,3)4/h1-9H,17H2,(H3,18,19)(H3,20,21);1H3,(H,2,3,4)/b4-1+;

InChI key

JYYHWWVHQKRPOO-DYVSEJHDSA-N

Application

suitable for the detection of DNA


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters