Skip to Content
Merck

Skip To

08491

D-Ribonic acid lithium salt

≥95.0% (TLC)

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C5H10O6 · xLi+
CAS Number:
Molecular Weight:
166.13 (free acid basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

assay

≥95.0% (TLC)

optical activity

[α]/D 6.5±1.0°, 24 hr, c = 1 in 1 M HCl

application(s)

clinical testing

storage temp.

2-8°C

SMILES string

OC[C@@H](O)[C@@H](O)[C@@H](O)C(O)=O.[Li+]

InChI

1S/C5H10O6.Li/c6-1-2(7)3(8)4(9)5(10)11;/h2-4,6-9H,1H2,(H,10,11);/q;+1/t2-,3-,4-;/m1./s1

InChI key

TVHSDAFFJAXPAS-WPFDICAFSA-N

Storage Class

11 - Combustible Solids

wgk

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
W212806-1KG-K04061837845482
381438-100ML04061831961072
226076-500G04061838779984
W212806-5KG-K04061837845499
W212806-SAMPLE-K04061837845505
55988-1ML04061832583266
226076-100G04061838779977
08491-10MG04061838652478
08491-250MG04061838652485
08491-50MG04061833414798

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service