Skip to Content
Merck

Skip To

07841

(R)-Mevalonic acid 5-phosphate lithium salt

95% (TLC)

Synonym(s):

(R)-3-Hydroxy-3-methyl-5-phosphonooxy-pentanoic acid lithium salt, (R)-5-Phosphomevalonate lithium salt, R-MVAP-Li

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Check Cart for Availability
€ 462,00
€ 346,50

About This Item

Empirical Formula (Hill Notation):
C6H13O7P · xLi+
CAS Number:
Molecular Weight:
228.14 (free acid basis)
UNSPSC Code:
12352106
NACRES:
NA.25
Assay:
95% (TLC)

€ 346,50

List Price€ 462,00Save 25%
Web-Only Promotion

Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

95% (TLC)

optical activity

[α]/D -3.2±0.5°, c = 0.1 in 0.2 M HCl

storage temp.

−20°C

SMILES string

[P](=O)([O-])([O-])OCC[C@@](O)(CC(=O)[O-])C

InChI

1S/C6H13O7P/c1-6(9,4-5(7)8)2-3-13-14(10,11)12/h9H,2-4H2,1H3,(H,7,8)(H2,10,11,12)/p-3/t6-/m1/s1

InChI key

OKZYCXHTTZZYSK-ZCFIWIBFSA-K

Application

Mevalonic acid 5-phosphate (M5P) may be used as a reference material in assays for the separation or detection of mevalonic acid 5-phosphate. Mevalonic acid 5-phosphate is the immediate precursor of isopentenyl pyrophosphate (IPP) and the mevalonate pathway which leads to isoprenoid biosynthesis. Mevalonic acid 5-phosphate may be used as a substrate to identify and characterize phosphomevalonate kinase(s).

Biochem/physiol Actions

Metabolite of the mevalonate pathway, which plays a key role in the biosynthesis of sterols, dolichol, heme and ubiquinone. Of interest for research in the disease areas oncology, autoimmune diseases, artherosclerosis and Alzheimer disease, as well as for inherited deficiencies of mevalonate kinase.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
067117984990469
assay

95% (TLC)

assay

≥95.0% (GC), 4-9% (Lithium, ICP)

assay

95% (TLC)

assay

≥96.0% (GC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

optical activity

[α]/D -3.2±0.5°, c = 0.1 in 0.2 M HCl

optical activity

[α]/D -19.0±2.0°, c = 0.5 in 1 M HCl

optical activity

-

optical activity

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters