Skip to Content
Merck
All Photos(1)

Key Documents

0711500

Sigma-Aldrich

Pentafluorophenyl benzoate

≥98.0% (HPLC)

Synonym(s):

Benzoic acid pentafluorophenyl ester, BzOPfp

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€411.00
100 G
€3,110.00

€411.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
10 G
€411.00
100 G
€3,110.00

About This Item

Linear Formula:
C6H5CO2C6F5
Molecular Weight:
288.17
Beilstein:
2299215
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.28

€411.00


Estimated to ship on31 May 2025


Request a Bulk Order

Assay

≥98.0% (HPLC)

mp

73-74 °C (lit.)

storage temp.

2-8°C

SMILES string

Fc1c(F)c(F)c(OC(=O)c2ccccc2)c(F)c1F
Fc1c(F)c(F)c(OC(=O)c2ccccc2)c(F)c1F

InChI

1S/C13H5F5O2/c14-7-8(15)10(17)12(11(18)9(7)16)20-13(19)6-4-2-1-3-5-6/h1-5H

InChI key

WZPWTXZSQHIABL-UHFFFAOYSA-N

Application

Active ester for the selective benzoylation of amino groups[1]

Other Notes

Currently available only in Europe.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Igolen, C. Morin
The Journal of Organic Chemistry, 45, 4802-4802 (1980)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service