Skip to Content
Merck
All Photos(1)

Documents

06857

Sigma-Aldrich

Nα-Benzoyl-Asn-Gly-Thr amide trifluoroacetate salt

≥90% (HPLC)

Synonym(s):

Nα-Benzoyl-L-asparaginylglycyl-L-threoninamide trifluoroacetate salt, Bz-Asn-Gly-Thr-NH2 trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23N5O6 · xC2HF3O2
CAS Number:
Molecular Weight:
393.39 (free base basis)
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Assay

≥90% (HPLC)

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.C[C@@H](O)[C@H](NC(=O)CNC(=O)[C@H](CC(N)=O)NC(=O)c1ccccc1)C(N)=O

InChI

1S/C17H23N5O6.C2HF3O2/c1-9(23)14(15(19)26)22-13(25)8-20-17(28)11(7-12(18)24)21-16(27)10-5-3-2-4-6-10;3-2(4,5)1(6)7/h2-6,9,11,14,23H,7-8H2,1H3,(H2,18,24)(H2,19,26)(H,20,28)(H,21,27)(H,22,25);(H,6,7)/t9-,11+,14+;/m1./s1

InChI key

FAAYXOTZFOVHSZ-YPMBYOOKSA-N

Application

Enzyme substrate for testing N-glycosylation.
Nα-Benzoyl-Asn-Gly-Thr amides can function as acceptor peptides for oligosaccharyltransferases and have been used to study their activity.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Dieckmann-Schuppert et al.
Biochimica et biophysica acta, 1199(1), 37-44 (1994-01-05)
Synthetic peptides were used to probe O- and N-glycosylation reactions in cell-free systems of the parasitic protozoa Plasmodium falciparum, Toxoplasma gondii, and Trypanosoma brucei brucei. O-Glycosylation of the peptide Pro-Tyr-Thr-Val-Val was observed with lysates from all organisms. However, the spectrum
E Bause et al.
The Biochemical journal, 312 ( Pt 3), 979-985 (1995-12-15)
Oligosaccharyltransferase (OST), an integral component of the endoplasmic-reticulum membrane, catalyses the transfer of dolichyl diphosphate-linked oligosaccharides to specific asparagine residues forming part of the Asn-Xaa-Thr/Ser sequence. We have studied the binding and catalytic properties of the enzyme from pig liver

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service