Skip to Content
Merck
All Photos(1)

Key Documents

03722

Sigma-Aldrich

Atto 580 Q

BioReagent

Synonym(s):

Atto 580 quencher, Atto Q 580

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€419.00

€419.00


Estimated to ship on27 April 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€419.00

About This Item

MDL number:
UNSPSC Code:
12352108
NACRES:
NA.32

€419.00


Estimated to ship on27 April 2025


Request a Bulk Order

product line

BioReagent

Quality Level

manufacturer/tradename

ATTO-TEC GmbH

technique(s)

UV/Vis spectroscopy: suitable

solubility

DMF: soluble
DMSO: soluble

suitability

corresponds for UV test

storage temp.

−20°C

General description

Atto 580Q is a new quencher for fluorescence characterized by high molecular absorption (110.000) and an absorption maximum at 586 nm (H2O). Atto 580Q is characterized by a high photostability and thermostability. It is moderately hydrophilic, and soluble in polar solvents as DMF or DMSO.

Application

Ato 580Q can be utilized as a fluorescence quencher (λ absorbance = 586 nm) on amine-labeled nucleotides for FRET experiments. Works well in combination with the following fluorescent dyes: TAMRA, ATTO 550, ATTO 565, ATTO 590, and ROX.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service