Skip to Content
Merck

03038

Tris(bathophenanthrolinedisulfonate)ruthenium(II) sodium salt solution

suitable for fluorescence, 20 mM in H2O

Synonym(s):

RuBPS, Rubin stain, Ruby stain, Ruthenium II tris (bathophenanthroline disulfonate), Ruthenium(II)-tris-(bathophenanthrolin-disulfonat) sodium salt solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C72H42N6Na4O18RuS6
CAS Number:
Molecular Weight:
1664.55
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12171500
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


shelf life

limited shelf life, expiry date on the label

concentration

20 mM in H2O

solubility

H2O: soluble

fluorescence

λex 445 nm; λem 615 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[Na+].[Na+].[Ru++].[O-]S(=O)(=O)c1ccc(cc1)-c2ccnc3c2ccc4c(ccnc34)-c5ccc(cc5)S([O-])(=O)=O.[O-]S(=O)(=O)c6ccc(cc6)-c7ccnc8c7ccc9c(ccnc89)-c%10ccc(cc%10)S([O-])(=O)=O.[O-]S(=O)(=O)c%11ccc(cc%11)-c%12ccnc%13c%12ccc%14c(ccnc%13%14)-c%15ccc(cc%15)S([O-])(=O)=O

InChI

1S/3C24H16N2O6S2.4Na.Ru/c3*27-33(28,29)17-5-1-15(2-6-17)19-11-13-25-23-21(19)9-10-22-20(12-14-26-24(22)23)16-3-7-18(8-4-16)34(30,31)32;;;;;/h3*1-14H,(H,27,28,29)(H,30,31,32);;;;;/q;;;4*+1;+2/p-6

InChI key

AXMFYAOBYBXMLC-UHFFFAOYSA-H

Application

Ruby Stain offers good sensitivity and minimal interference with MALDI-TOF analyis. It has been demonstrated as an effective fluorescent stain for the analysis of 2D-gel separated proteins .
Used for the detection of DNA

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.
One package includes 50 μl concentrate. Dilution into 1 L water yields a 1 μM solution

Analysis Note

The dye can be excited with standard light sources. Excitation with 300 nm (UV light) or 473 nm (blue laser) is suitable.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library