Skip to Content
Merck
All Photos(1)

Key Documents

F200

Sigma-Aldrich

Fuller′s earth

-100 mesh particle size

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52
Pricing and availability is not currently available.

form

powder

Quality Level

particle size

-100 mesh

Looking for similar products? Visit Product Comparison Guide

Application


  • Kinetic and equilibrium profile of the adsorptive removal of Acid Red 17 dye by surfactant-modified fuller′s earth.: This study explores the adsorption capabilities of surfactant-modified fullers earth for removing Acid Red 17 dye from aqueous solutions, detailing the kinetic and equilibrium aspects of the adsorption process (Shah et al., 2017).

  • Efficacy of scalp hair decontamination following exposure to vapours of sulphur mustard simulants 2-chloroethyl ethyl sulphide and methyl salicylate.: Investigates the effectiveness of fullers earth in decontaminating scalp hair exposed to chemical vapors, demonstrating its potential use in chemical hazard mitigation (Spiandore et al., 2017).

  • In vitro skin decontamination of the organophosphorus pesticide Paraoxon with nanometric cerium oxide CeO(2).: While primarily focusing on nanometric cerium oxide, this study references fullers earth as part of comparative decontamination methodologies (Salerno et al., 2017).

  • Production and characterization of refined oils obtained from Indian oil sardine (Sardinella longiceps).: Discusses the use of fullers earth in the refining process of fish oils, highlighting its application in the food industry for oil purification (Chakraborty and Joseph, 2015).

  • Preparation of methacrylic acid-modified rice husk improved by an experimental design and application for paraquat adsorption.: Although the primary focus is on modified rice husk, this paper also examines fullers earth for paraquat adsorption, offering insights into its use in environmental remediation (Hsu et al., 2009).

Features and Benefits

Processed to increase absorption capacity and reduce volatiles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juanjuan Zhang et al.
3 Biotech, 10(1), 12-12 (2019-12-26)
The present work showed the preparation of highly stable spherical nanobubbles using 3 mg/ml of soy lipid and 1% of Tween 80 as surfactant. The prepared nanobubbles were characterized using TEM and zeta-potential analyzer, which confirmed the formation of spherical nanobubbles
Youjin Kim et al.
Polymers, 12(9) (2020-09-03)
In this study, hybrid fillers of spherically shaped aggregated boron nitride (a-BN) attached with SAC305, were fabricated via simple stirring and the vacuum filtration method. a-BN was used as the primary conductive filler incorporated with epoxy resin, and these fillers
A new method for sequential extraction of oxytocin and antidiuretic hormone for radioimmunoassay.
R P Sequeira et al.
Indian journal of experimental biology, 22(4), 215-219 (1984-04-01)
Untractable fecalith's and hypercalcemia, both associated with Fuller's earth therapy in fatal case of paraquat poisoning.
J Vandenbogaerde et al.
Journal of toxicology. Clinical toxicology, 19(9), 1011-1012 (1982-11-01)
S R Drachman et al.
Solid state nuclear magnetic resonance, 9(2-4), 257-267 (1998-02-27)
Fuller's Earth, a dioctahedral calcium montmorillonite clay mineral of the smectite group, undergoes thermal transformation via a series of complex intermediate states that are highly structurally disordered. Fuller's Earth is a commercially significant material with considerable levels of paramagnetic iron

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service