Skip to Content
Merck
All Photos(1)

Key Documents

71361

Sigma-Aldrich

Sodium carbonate hydrate

purum p.a., ≥99.0% (calc. based on dry substance, T), 9-10 mol/mol water

Synonym(s):

Soda

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CO3 · aq
CAS Number:
Molecular Weight:
105.99 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
form:
crystals and lumps
fine crystals
solubility:
water: soluble 142.3 g/L at 20 °C

grade

purum p.a.

Assay

≥99.0% (calc. based on dry substance, T)

form

crystals and lumps
fine crystals

impurities

9-10 mol/mol water

pH

12 (25 °C, 143 g/L)

solubility

water: soluble 142.3 g/L at 20 °C

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Na+].[Na+].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-]C([O-])=O

InChI

1S/CH2O3.2Na.10H2O/c2-1(3)4;;;;;;;;;;;;/h(H2,2,3,4);;;10*1H2/q;2*+1;;;;;;;;;;/p-2

InChI key

XYQRXRFVKUPBQN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium carbonate decahydrate is a hydrated form of Na2CO3. IR spectral investigations suggest its spectra is similar to the heptahydrate form. It undergoes thermal dehydration to monohydrate form in a batch, fluidized bed reactor.

Application

Sodium carbonate decahydrate may be used to investigate the hydrolysis of poly(β-cyclodextrin-co-citric acid) (CDP, water-insoluble polymer). Polymeric hydrolysates were investigated by HPLC.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahdiyeh Otoukesh et al.
Food chemistry, 321, 126720-126720 (2020-04-11)
Migration of N,N-Bis(2-hydroxyethyl) alkyl(C8-C18)amines from five different polypropylene capsules to Tenax® and coffee powder have been studied. A single step extraction-cleanup procedure using salting out liquid-liquid extraction (SALLE) method followed by ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) was applied.
Dehydration of sodium carbonate decahydrate to monohydrate in a fluidized bed.
Hartman M, et al.
AIChE Journal, 47(10), 2333-2333 (2001)
Chemical structure of poly (β-cyclodextrin-co-citric acid).
Bednarz S, et al.
Journal of Applied Polymer Science, 119(6), 3511-3520 (2011)
An infra-red study of the hydrates of sodium carbonate.
Buijs K and Schutte CJH.
Spectrochimica Acta Part A: Molecular Spectroscopy, 17(9), 917-920 (1961)
Hongje Jang et al.
ACS nano, 9(3), 2696-2703 (2015-01-07)
The polyvinylpyrrolidone (PVP)-coated spherically clustered porous gold-silver alloy nanoparticle (PVP-SPAN) was prepared by low temperature mediated, partially inhibited galvanic replacement reaction followed by silver etching process. The prepared porous nanostructures exhibited excellent photothermal conversion efficiency under irradiation of near-infrared light

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service