Skip to Content
Merck
All Photos(3)

Key Documents

398063

Sigma-Aldrich

Sodium dichromate dihydrate

ACS reagent, ≥99.5%

Synonym(s):

Sodium bichromate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2Cr2O7 · 2H2O
CAS Number:
Molecular Weight:
298.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
crystals
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Assay

≥99.5%

form

crystals

reaction suitability

reagent type: oxidant

impurities

≤0.005% NH4OH ppt.
≤0.005% insolubles

pH

3.5 (20 °C, 100 g/L)

mp

91 °C (lit.)

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.01%

cation traces

Al: ≤0.002%
Ca: ≤0.003%

SMILES string

O.O.[Na+].[Na+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

InChI

1S/2Cr.2Na.2H2O.7O/h;;;;2*1H2;;;;;;;/q;;2*+1;;;;;;;;2*-1

InChI key

JYDRNIYTFCBIFC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium dichromate dehydrate (SDD), a hexavalent chromium compound[1] is commonly used as an oxidizing agent.[2] Chromium (VI) compounds are genotoxic and group 1 IARC (International Agency for Research on Cancer) carcinogen.[3] SDD is one of the common contaminants found in drinking water. Its toxic effects and carcinogenicity has been studied.[1][3][4][5]

Application

Sodium dichromate dehydrate may be used in preparing Jones reagent which is employed for oxidation reactions.[6] It may be used in the oxidation of arylalkanes.[7]

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 1B - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1 Inhalation

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaken not stirred; oxidation of alcohols with sodium dichromate.
Lou JD, et al.
Tetrahedron Letters, 47(3), 311-313 (2006)
Chromium Oxidations in Organic Chemistry
Cainelli G and Cardillo G.
Chromium Oxidations in Organic Chemistry, 29-32G-29-32G (2012)
Lack of genotoxic effects in hematopoietic and gastrointestinal cells of mice receiving chromium (VI) with the drinking water.
De Flora S, et al.
Mutation Research, Reviews in Mutation Research, 659(1), 60-67 (2008)
Synthesis of novel types of polyester glycodendrimers as potential inhibitors of urinary tract infections.
Jean-d'Amour KT, et al.
New. J. Chem., 39(5), 4115-4127 (2015)
A chronic oral reference dose for hexavalent chromium-induced intestinal cancer.
Thompson CM, et al.
Journal of Applied Toxicology, 34(5), 525-536 (2014)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service