Skip to Content
Merck
All Photos(2)

Key Documents

358983

Sigma-Aldrich

Ammonium sulfite monohydrate

92%

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€47.80

€47.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
€47.80

About This Item

Linear Formula:
(NH4)2SO3 · H2O
CAS Number:
Molecular Weight:
134.16
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.55
grade:
for analytical purposes
form:
crystals

€47.80


Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

Quality Level

Assay

92%

form

crystals

density

1.41 g/mL at 25 °C (lit.)

SMILES string

N.N.O.OS(O)=O

InChI

1S/2H3N.H2O3S.H2O/c;;1-4(2)3;/h2*1H3;(H2,1,2,3);1H2

InChI key

ASPXBYAQZVXSNS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ammonium sulfite monohydrate is generally used as a reducing agent and sulfonating agent in the organic synthesis.

Application

Ammonium sulfite monohydrate is used in the wafers cleaning, etching, and silicon modification procedures.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yan B Vogel et al.
Nature communications, 8(1), 2066-2066 (2017-12-14)
Predicting or manipulating charge-transfer at semiconductor interfaces, from molecular electronics to energy conversion, relies on knowledge generated from a kinetic analysis of the electrode process, as provided by cyclic voltammetry. Scientists and engineers encountering non-ideal shapes and positions in voltammograms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service