343005
Iron(III) oxide
pieces, 3-12 mm, 99.8% trace metals basis
Synonym(s):
Ferric oxide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
Fe2O3
CAS Number:
Molecular Weight:
159.69
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
form:
pieces
sintered
sintered
Assay
99.8% trace metals basis
form
pieces
sintered
particle size
3-12 mm
SMILES string
O=[Fe]O[Fe]=O
InChI
1S/2Fe.3O
InChI key
JEIPFZHSYJVQDO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Suitable for vacuum deposition.
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
William B Ingler et al.
Journal of the American Chemical Society, 126(33), 10238-10239 (2004-08-19)
Stable zinc-doped iron(III) oxide thin films that exhibit p-type behavior were synthesized by spray pyrolytic deposition (SPD) on conducting indium-doped tin oxide-coated glass substrate. The highest photocurrent density of 1.1 mA/cm2 was observed at an illumination intensity of 40 mW/cm2
Synthesis and photoelectrochemical properties of nanoporous iron (III) oxide by potentiostatic anodization.
Prakasam HE, et al.
Nanotechnology, 17(17), 4285-4285 (2006)
The structure of iron (III) oxide-hydrate gels.
Van der Giessen AA.
J. Inorg. Nucl. Chem., 28(10), 2155-2159 (1966)
Light-induced reduction of natural iron (III) oxide and its relevance to phytoplankton.
Finden DAS, et al.
Nature, 309, 783-784 (1984)
Electrochemistry and photoelectrochemistry of iron (III) oxide.
Dare-Edwards MP, et al.
J. Chem. Soc., Faraday, 79(9), 2027-2041 (1983)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service