Skip to Content
Merck
All Photos(3)

Documents

31252

Sigma-Aldrich

Potassium cyanide

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCN
CAS Number:
Molecular Weight:
65.12
Beilstein:
3593645
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent
puriss. p.a.

Agency

ISO
USP/NF
reag. Ph. Eur.

Assay

≥97.0%

form

crystals

impurities

≤0.01% thiocyanate (SCN)

pH

11.5 (20 °C, 20 g/L)

mp

634 °C (lit.)

anion traces

chloride (Cl-): ≤200 mg/kg
phosphate (PO43-): ≤50 mg/kg
sulfate (SO42-): ≤400 mg/kg
sulfide (S2-): ≤5 mg/kg

cation traces

Fe: ≤100 mg/kg
Na: ≤5000 mg/kg
Pb: ≤2 mg/kg

SMILES string

[K]C#N

InChI

1S/CN.K/c1-2;

InChI key

YUZRZFQHUCKACF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassium cyanide, a potassium salt, is a colorless hygroscopic compound.

Application

Potassium cyanide has been used:
  • in the reduction of cassiterite for Sn isotope analysis
  • gold microelectrode fabrication
  • to prepare the calibration curve for colorimetric determination of hydrocyanic acid (HCN) levels in Sorghum and Sudangrass species

Biochem/physiol Actions

Potassium cyanide (KCN) is useful in dyeing, printing, photography, metal cleaning, and manufacturing. It is considered a reversible cytochrome oxidase inhibitor that blocks cellular respiration. KCN is poisonous when inhaled or ingested.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Met. Corr. 1 - STOT RE 1

Target Organs

Thyroid

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and measurement of cassiterite for Sn isotope analysis
Mathur R, et al.
Geostandards and Geoanalytical Research, 41(4), 701-707 (2017)
Cyano compounds, inorganic
Gail E, et al.
Ullmann's Encyclopedia of Industrial Chemistry (2000)
Monochloramine-sensitive amperometric microelectrode: optimization of gold, platinum, and carbon fiber sensing materials for removal of dissolved oxygen interference
Lee WH, et al.
Ionics, 21(9), 2663-2674 (2015)
The effects of different sowing time and harvesting height on hydrocyanic acid content in some silage sorghum (sorghum bicolor l.) varieties
Mehmet
Turkish journal Of Field Crops, 22(2), 211-217 (2017)
Chemical Agents
Responding to Terrorism, 233-344 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service