Skip to Content
Merck
All Photos(1)

Key Documents

308269

Supelco

2-Methoxyethyl acetate

suitable for HPLC, ≥99%

Synonym(s):

1-Acetoxy-2-methoxyethane, Ethylene glycol monomethyl ether acetate, Methyl Cellosolve® acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH2OCH3
CAS Number:
Molecular Weight:
118.13
Beilstein:
1700761
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21
grade:
HPLC grade
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
145 °C (lit.)
Pricing and availability is not currently available.

grade

HPLC grade

Assay

≥99%

form

liquid

purified by

glass distillation

expl. lim.

0.34-6.3 %

technique(s)

HPLC: suitable

impurities

<0.050% water

evapn. residue

<0.0005%

refractive index

n20/D 1.402 (lit.)

bp

145 °C (lit.)

mp

−65 °C (lit.)

density

1.009 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 254 nm Amax: 1.00
λ: 275 nm Amax: 0.15
λ: 300 nm Amax: 0.05
λ: 350-400 nm Amax: 0.01

application(s)

food and beverages

SMILES string

COCCOC(C)=O

InChI

1S/C5H10O3/c1-5(6)8-4-3-7-2/h3-4H2,1-2H3

InChI key

XLLIQLLCWZCATF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Cellosolve is a registered trademark of Union Carbide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Smialowicz et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 18(4), 621-627 (1992-05-01)
Oral dosing of adult male F344 rats with the glycol ether 2-methoxyethanol (ME) or its principal metabolite 2-methoxyacetic acid (MAA) results in the suppression of the primary plaque-forming cell (PFC) response to trinitrophenyl-lipopolysaccharide (TNP-LPS). In the present study, the PFC
G Johanson
Critical reviews in toxicology, 30(3), 307-345 (2000-06-14)
Ethylene glycol monomethyl ether (EGME) and its acetate ester (EGMEA) are highly flammable, colorless, moderately volatile liquids with very good solubility properties. They are used in paints, lacquers, stains, inks and surface coatings, silk-screen printing, photographic and photo lithographic processes
T-S Shih et al.
Occupational and environmental medicine, 60(2), 130-135 (2003-01-30)
To examine the association between 2-methoxyethanol (2-ME) exposure and haematological effects, as well as the recovery from these haematological effects with continuous reduction in exposure to 2-ME. Twenty nine exposed and 90 non-exposed workers were recruited. Haematological parameters, eight hour
[Hygienic regulation of ethylene glycol monomethacrylic ether in reservoir water].
I I Datsenko et al.
Gigiena i sanitariia, (9)(9), 75-76 (1989-09-01)
Hong-I Chen et al.
Journal of occupational health, 49(4), 285-293 (2007-08-11)
The objective of this study was to evaluate an intervention program, implemented in year 1999, of wearing rubber gloves in addition to engineering control, and to follow-up the hematological effects of 2-ethoxyethanol acetate (2-EEA) exposure among workers in a silk-screening

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service