Skip to Content
Merck
All Photos(1)

Documents

20911

Sigma-Aldrich

Cadmium nitrate tetrahydrate

purum p.a., ≥99.0% (T)

Synonym(s):

Nitric acid, cadmium salt tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cd(NO3)2 · 4H2O
CAS Number:
Molecular Weight:
308.48
Beilstein:
8156730
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

purum p.a.

Assay

≥99.0% (T)

form

powder or crystals

mp

59.5 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-][N+](=O)O[Cd]O[N+]([O-])=O

InChI

1S/Cd.2NO3.4H2O/c;2*2-1(3)4;;;;/h;;;4*1H2/q+2;2*-1;;;;

InChI key

QOYRNHQSZSCVOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 1B - STOT RE 1

Target Organs

Kidney,Bone

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guiqiu Chen et al.
Colloids and surfaces. B, Biointerfaces, 117, 199-205 (2014-03-19)
This study details a novel method for the extracellular microbial synthesis of cadmium sulfide (CdS) quantum dots (QDs) by the white rot fungus Phanerochaete chrysosporium. P. chrysosporium was incubated in a solution containing cadmium nitrate tetrahydrate, which became yellow from
Hui-Ting Wang et al.
Acta crystallographica. Section C, Structural chemistry, 71(Pt 7), 549-553 (2015-07-07)
A new cadmium-thiocyanate complex, namely catena-poly[1-carboxymethyl-4-(dimethylamino)pyridinium [cadmium(II)-tri-μ-thiocyanato-κ(4)N:S;κ(2)S:N] [[[4-(dimethylamino)pyridinium-1-acetate-κ(2)O,O']cadmium(II)]-di-μ-thiocyanato-κ(2)N:S;κ(2)S:N]], {(C9H13N2O2)[Cd(NCS)3][Cd(NCS)2(C9H12N2O2)]}n, was synthesized by the reaction of 4-(dimethylamino)pyridinium-1-acetate, cadmium nitrate tetrahydrate and potassium thiocyanide in aqueous solution. In the crystal structure, two types of Cd(II) atoms are observed in distorted octahedral coordination
Qiang Li et al.
Acta crystallographica. Section C, Structural chemistry, 70(Pt 11), 1054-1056 (2014-11-06)
A new cadmium dicyanamide complex, poly[tetramethylphosphonium [μ-chlorido-di-μ-dicyanamido-κ(4)N(1):N(5)-cadmium(II)]], [(CH3)4P][Cd(NCNCN)2Cl], was synthesized by the reaction of tetramethylphosphonium chloride, cadmium nitrate tetrahydrate and sodium dicyanamide in aqueous solution. In the crystal structure, each Cd(II) atom is octahedrally coordinated by four terminal N atoms
Xue-Mei Li et al.
Journal of inorganic biochemistry, 101(8), 1165-1171 (2007-06-23)
A novel hybridization indicator, bis(benzimidazole)cadmium(II) dinitrate (Cd(bzim)(2)(NO(3))(2)), was utilized to develop an electrochemical DNA biosensor for the detection of a short DNA sequence related to the hepatitis B virus (HBV). The sensor relies on the immobilization and hybridization of the
Francesco Crea et al.
Annali di chimica, 95(11-12), 767-778 (2006-01-10)
In order to continue the investigation on the formation of hetero-metal polynuclear hydrolytic species, in this paper we report some results (at I = 0.16 mol L(-1) in NaNO3, at t = 25 degrees C by potentiometry, ISE-H+, glass electrode)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service