Skip to Content
Merck
All Photos(1)

Key Documents

Z804339

HPLC Qualification Kit

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.21

grade

certified reference material

manufacturer/tradename

Jaytee Biosciences Ltd (CRM-1100)

technique(s)

HPLC: suitable

General description

CRM for HPLC.

Contains qualification standards for

  • UV Multi Wavelength
  • Low Wavelength
  • Absorbance Linearity
  • HPLC Autosampler

This HPLC qualification kit has been developed to be used in any laboratory regardless of the regulator and are as relevant to a pharmaceutical company as to a contract lab working to ISO 17025.

All products come supplied in flame-sealed ampoules to increase the shelf life and prevent contamination, with full certification.

Traceable instrument qualification adds confidence to all data generated. In addition, your audit trail is backed up by the independent verification by UKAS to ISO 17034.

For US please order the products included in the kit separately: Z803898 (HPLC Autosampler Standards), Z804002 (UV Absorbance Linearity Standards), Z804126 (UV Low Wavelength Standards), Z804223 (UV Multi Wavelength Standards).

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 1

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service