Key Documents
Z531448
Sigma-Aldrich
Aldrich® Graham condenser
Overall H 400 mm, joint: ST/NS 24/40
Select a Size
1 EA
€230.00
€230.00
Estimated to ship on15 April 2025
Select a Size
Change View
1 EA
€230.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
€230.00
Estimated to ship on15 April 2025
Joint
joint: ST/NS24/40
availability
not available in EU
Overall H
400 mm
Related Categories
General description
The Graham condenser has a large cooling surface area by allowing coolant to circulate around the inner coil. Condensers have a standard taper outer joint and a lower inner drip-tip joint.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)