Skip to Content
Merck
All Photos(1)

Key Documents

Y0001601

Alfacalcidol for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1α-Hydroxyvitamin D3, 1α-Hydroxycholecalciferol, Alfacalcidol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O2
CAS Number:
Molecular Weight:
400.64
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C3\C[C@@H](O)C[C@H](O)C3=C

InChI

1S/C27H44O2/c1-18(2)8-6-9-19(3)24-13-14-25-21(10-7-15-27(24,25)5)11-12-22-16-23(28)17-26(29)20(22)4/h11-12,18-19,23-26,28-29H,4,6-10,13-17H2,1-3,5H3/b21-11+,22-12-/t19-,23-,24-,25+,26+,27-/m1/s1

InChI key

OFHCOWSQAMBJIW-AVJTYSNKSA-N

Gene Information

human ... VDR(7421)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

This European Pharmacopoeia reference standard is intended for use only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Synthetic vitamin D3 analog that is metabolized to 1,25-dihydroxycholecalciferol. Inhibits bone resorption and stimulates bone formation. Supresses parathyroid hormone secretion by bovine parathyroid cells.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 1 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vitamin D and metabolites measurement by tandem mass spectrometry
van den Ouweland JMW, et al.
Reviews in Endocrine & Metabolic Disorders, 14(2), 159-184 (2013)
Siobhan O'Donnell et al.
Journal of bone and mineral metabolism, 26(6), 531-542 (2008-11-04)
Our objective was to conduct a systematic review on the benefits and harms of calcitriol and alfacalcidol in the reduction of fracture and fall risk. Randomized controlled trials (RCTs) comparing these agents to placebo or calcium and reporting fracture and
Noboru Kubodera
Mini reviews in medicinal chemistry, 9(12), 1416-1422 (2009-11-26)
Many readers may have only a vague idea about vitamin D. This is made complicated, in part, because it is also expressed with suffixes such as vitamin D(2) or vitamin D(3). Otherwise the prefix of "active" is also occasionally used.
P Sambrook
Calcified tissue international, 65(4), 341-343 (1999-09-15)
There is increasing evidence that D-hormones (active vitamin D metabolites) are effective agents in preventing posttransplantation bone loss of various organs. This effect may relate to several mechanisms including reversal of secondary hyperparathyroidism from cyclosporine, reversal of corticosteroid effects on
Florent Richy et al.
Aging clinical and experimental research, 17(2), 133-142 (2005-06-28)
Alfacalcidol (1-alpha-hydroxyvitamin D3) is a non-endogenous analog of vitamin D which can bypass the renal and intestinal regulatory mechanisms that control the production of calcitriol (1,25-hydroxyvitamin D3, the active form of vitamin D, D-Hormone). Alfacalcidol may be metabolized into calcitriol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service