Skip to Content
Merck
All Photos(1)

Key Documents

Y0001324

Sucralfate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Sucrose octasulfate–aluminum complex, Sucralfate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

sucralfate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O[Al+]O.O[Al+]O.O[Al+]O.O[Al+]O.O[Al+]O.O[Al+]O.O[Al+]O.O[Al+]O.O[Al](O)O.O[Al](O)O.O[Al](O)O.O[Al](O)O.O[Al](O)O.O[Al](O)O.O[Al](O)O.O[Al](O)O.[O-]S(=O)(=O)OC[C@H]1O[C@@H](O[C@]2(COS([O-])(=O)=O)O[C@H](OS([O-])(=O)=O)[C@@H](OS([O-])(=O)=O)[C@@H]2OS([O-])(=O)=O)[C@H](OS([O-])(=O)=O)[C@@H](OS([O-])(=O)=O)[C@@H]1OS([O-])(=O)=O

InChI

1S/C11H20O35S8.16Al.40H2O/c12-47(13,14)36-1-3-4(41-49(18,19)20)5(42-50(21,22)23)6(43-51(24,25)26)9(38-3)39-11(2-37-48(15,16)17)8(45-53(30,31)32)7(44-52(27,28)29)10(40-11)46-54(33,34)35;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h3-10H,1-2H2,(H,12,13,14)(H,15,16,17)(H,18,19,20)(H,21,22,23)(H,24,25,26)(H,27,28,29)(H,30,31,32)(H,33,34,35);;;;;;;;;;;;;;;;;40*1H2/q;16*+3;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/p-48/t3-,4-,5+,6-,7+,8+,9+,10-,11-;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/m1......................................................../s1

InChI key

JTZPPHUZZDKEOC-RBQAPOGLSA-A

Gene Information

human ... PGA5(5222)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sucralfate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S K Lam et al.
Scandinavian journal of gastroenterology. Supplement, 210, 89-91 (1995-01-01)
Sucralfate monotherapy has been shown to suppress but not eradicate Helicobacter pylori. The combination of sucralfate with antibiotic(s) has been evaluated recently in the treatment of H. pylori-positive duodenal ulcer. The aim of this article is to review the efficacy
B A Hemstreet
The Annals of pharmacotherapy, 35(3), 360-364 (2001-03-23)
To assess the potential for the development of aluminum toxicity in patients with renal insufficiency or chronic renal failure who are taking sucralfate. Clinical literature accessed through MEDLINE (1966-December 1999) and International Pharmaceutical Abstracts (1970-December 1999). Key search terms included
I N Marks et al.
The American journal of medicine, 91(2A), 95S-101S (1991-08-08)
Controversial aspects relating to acid secretory changes following duodenal ulcer healing, and the effect of histamine-2 (H2)-receptor blockers on such changes are discussed. Recent studies comparing the effects of ranitidine or sucralfate on changes in the acid secretory responses in
Sucralfate in clinical practice.
S K Lam et al.
Journal of gastroenterology and hepatology, 9(4), 401-411 (1994-07-01)
W D Rees
Scandinavian journal of gastroenterology. Supplement, 191, 4-6 (1992-01-01)
Sucralfate has a complex effect on the luminal and mucosal environment of the stomach and duodenum. Some of the actions are important in ulcer healing whilst others are important in preventing subsequent ulcer relapse. Although sucralfate has little direct effect

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service