Skip to Content
Merck
All Photos(1)

Key Documents

Y0001150

Dextropropoxyphene for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Dextropropoxyphene hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29NO2 · HCl
CAS Number:
Molecular Weight:
375.93
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dextropropoxyphene

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Cl-].N(C[C@H]([C@@](OC(=O)CC)(Cc2ccccc2)c1ccccc1)C)(C)C.[H+]

InChI

1S/C22H29NO2.ClH/c1-5-21(24)25-22(18(2)17-23(3)4,20-14-10-7-11-15-20)16-19-12-8-6-9-13-19;/h6-15,18H,5,16-17H2,1-4H3;1H/t18-,22+;/m1./s1

InChI key

QMQBBUPJKANITL-MYXGOWFTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dextropropoxyphene for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

India urged to reverse dextropropoxyphene ban.
Dinesh C Sharma
The Lancet. Oncology, 14(9), e344-e344 (2013-09-24)
Wayne A Ray et al.
Pharmacoepidemiology and drug safety, 22(4), 403-412 (2013-02-15)
The opioid analgesic propoxyphene was withdrawn from the US market in 2010, motivated by concerns regarding fatality in overdose and adverse cardiac effects, including prolongation of the QT interval. These concerns were based on case reports, summary vital statistics, and
Feng-Sheng Lin et al.
Acta anaesthesiologica Taiwanica : official journal of the Taiwan Society of Anesthesiologists, 50(2), 49-53 (2012-07-10)
Weak opioid combined with acetaminophen (APAP) has been proven to provide better analgesic efficacy and cause fewer complications than either drug alone. However, there are questions about whether different opioids, tramadol and propoxyphene, provide similar efficacy or safety. Thus, we
Cindy Bourne et al.
British journal of clinical pharmacology, 75(4), 1063-1067 (2012-09-05)
The risk of hypoglycaemia with tramadol (TRM) is not well described. Our aim was to analyze spontaneous reports of hypoglycaemia registered in the French Pharmacovigilance database and to compare these data with two other step-2 analgesic drugs. Cases of hypoglycaemia
Joseph L Schuller et al.
Expert opinion on pharmacotherapy, 13(13), 1825-1827 (2012-07-10)
The most commonly prescribed class of medications in the United States for chronic severe pain is opioid analgesics. Due to their low cost and widespread availability, the synthetic opioids are popular choices among clinicians and patients. However, there is an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service